Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 8
This intronless gene encodes a 70kDa heat shock protein which is a member of the heat shock protein 70 family. In conjuction with other heat shock proteins, this protein stabilizes existing proteins against aggregation and mediates the folding of newly translated proteins in the cytosol and in organelles. It is also involved in the ubiquitin-proteasome pathway through interaction with the AU-rich element RNA-binding protein 1. The gene is located in the major histocompatibility complex class III region, in a cluster with two closely related genes which encode similar proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 70 kda heat shock protein-2; chaperone; dnaK-type molecular chaperone HSP70-1; epididymis secretory protein Li 103; heat shock 70 kD protein 2; Heat shock 70 kDa protein 1; heat shock 70 kDa protein 1/2; Heat shock 70 kDa protein 1A; heat shock 70 kDa protein 1A/1B; Heat shock 70 kDa protein 1B; Heat shock 70 kDa protein 2; heat shock 70kD protein 1A; heat shock 70kDa protein 1A; heat shock 70kDa protein 1B; Heat shock protein family A member 1A; Heat shock protein family A member 1B; heat shock-induced protein; heat-shock 70-kilodalton protein 1B; HSP70-1; HSP70-1/HSP70-2; HSP70-2; HSP70.1; HSP70.1/HSP70.2; HSP70.2
Gene Aliases: BOS_21593; HEL-S-103; HSP70-1; HSP70-1A; HSP70-2; HSP70.1; HSP70I; HSP72; HSPA1; HSPA1A; HSPA1B; HSPA2; HSX70
UniProt ID: (Human) P0DMV8, (Bovine) Q27975
Entrez Gene ID: (Human) 3303, (Bovine) 282254
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support