Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This antibody well recognized the endogenous HSPA4 protein in testis and multiple cell lines.
Immunogen sequence: LYEGVDFYT SITRARFEEL NADLFRGTLE PVEKALRDAK LDKGQIQEIV LVGGSTRIPK IQKLLQDFFN GKELNKSINP DEAVAYGAAV QAAILIGDKS ENVQDLLLLD VTPLSLGIET AGGVMTPLIK RNTTIPTKQT QTFTTYSDNQ SSVLVQVYEG ERAMTKDNNL LGKFDLTGIP PAPRGVPQIE VTFDIDANGI LNVTAADKST GKENKITITN DKGRLSKDDI DRMVQEAERY KSEDEANRDR VAAKNALESY TYNIKQTVED EKLRGKISEQ DKNKILDKCQ EVINWLDRNQ MAEKDEYEHK QKELERVCNP IISKLYQGGP GGGSGGGGSG ASGGPTIEEV D (290-639 aa encoded by BC036107)
In cooperation with other chaperones, Hsp70s stabilize preexistent proteins against aggregation and mediate the folding of newly translated polypeptides in the cytosol as well as within organelles. These chaperones participate in all these processes through their ability to recognize nonnative conformations of other proteins. They bind extended peptide segments with a net hydrophobic character exposed by polypeptides during translation and membrane translocation, or following stress-induced damage.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Heat shock 70 kDa protein 2; heat shock 70kD protein 2; heat shock 70kDa protein 2; Heat shock protein; Heat shock protein 70.2; heat shock protein alpha 2; Heat shock protein family A member 2; heat shock protein, 70 kDa 2; Heat shock-related 70 kDa protein 2; HSP; HST; Testis-specific heat shock protein-related; testis-specific heat shock protein-related gene hst70
Gene Aliases: 70kDa; Hcp70.2; HSP70-2; HSP70-3; HSP70.2; HSP70A2; HSPA2; Hspt70; Hst70
UniProt ID: (Human) P54652, (Rat) P14659, (Mouse) P17156
Entrez Gene ID: (Human) 3306, (Rat) 60460, (Mouse) 15512
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support