Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MSGRSVPHA HPATAEYEFA NPSRLGEQRF GEGLLPEEIL TPTLYHGYYV RPRAAPAGEG SRAGASELRL SEGKFQAFLD VSHFTPDEVT VRTVDNLLEV SARHPQRLDR HGFVSREFCR TYVLPADVDP WRVRAALSHD GILNLEAPRG GRHLDTEVNE VYISLLPAPP DPEEEEEAAI VEP (1-182 aa encoded by BC109393)
The protein encoded by this gene belongs to the superfamily of small heat-shock proteins containing a conservative alpha-crystallin domain at the C-terminal part of the molecule. The protein is expressed preferentially in the heart and skeletal muscle. This protein regulates Myotonic Dystrophy Protein Kinase, which plays an important role in maintenance of muscle structure and function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DMPK-binding protein; heat shock 27kD protein 2; heat shock 27kDa protein 2; Heat shock protein; heat shock protein 2; heat shock protein beta 2; Heat shock protein beta-2; Heat shock protein family B member 2; heat-shock protein beta-2; HSP; HspB2; MGC133245; MKBP
Gene Aliases: Hs.78846; HSP27; HSPB2; LOH11CR1K; MKBP
UniProt ID: (Human) Q16082, (Rat) O35878
Entrez Gene ID: (Human) 3316, (Rat) 161476
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support