Search Thermo Fisher Scientific
Progen
Reconstitute lyophilized product with 1 mL distilled water.
A1C5 reacts with an epitope present on the N-terminus (between aa 15 and 35) of the nucleocapsid protein (S segment) of hantavirus CG 18-20 strain (Puumala serotype) and also cross-reacts with SNV (Sin Nombre virus) rN protein.