Product References
Molecular Dynamics Simulations Reveal Novel Interacting Regions of Human Prion Protein to Brucella abortus Hsp60 Protein.
Molecular biotechnology
Le-Dao HA,Dinh TT,Tran TL,Lee VS,Tran-Van H
Mon Apr 01 00:00:00 UTC 2024
Oxidation-Induced Protein Cross-Linking in Mammalian Cells.
ACS synthetic biology
Li H,Lv L,Tang S,Zang Y,Wan T,Wang D,Cai L,Ye H,Tan R,Wang N
Fri Apr 21 00:00:00 UTC 2023
Genetically encoded chemical crosslinking of carbohydrate.
Nature chemistry
Li S,Wang N,Yu B,Sun W,Wang L
Sun Jan 01 00:00:00 UTC 2023
Phosphoglycerate dehydrogenase activates PKM2 to phosphorylate histone H3T11 and attenuate cellular senescence.
Nature communications
Wu Y,Tang L,Huang H,Yu Q,Hu B,Wang G,Ge F,Yin T,Li S,Yu X
Fri Mar 10 00:00:00 UTC 2023
Escherichia coli has robust regulatory mechanisms against elevated peptidoglycan cleavage by lytic transglycosylases.
The Journal of biological chemistry
Liang Y,Zhao Y,Kwan JMC,Wang Y,Qiao Y
Sat Apr 01 00:00:00 UTC 2023
Vitamin B6 regulates IL-33 homeostasis to alleviate type 2 inflammation.
Cellular & molecular immunology
Zhu S,Zhong S,Cheng K,Zhang LS,Bai JW,Cao Z,Wang S,Chen W,Cheng S,Ma L,Ling Z,Huang Y,Gu W,Sun X,Yi C,Zhao M,Liang S,Xu JF,Sun B,Zhang Y
Sat Jul 01 00:00:00 UTC 2023
Strong immune responses and protection of PcrV and OprF-I mRNA vaccine candidates against Pseudomonas aeruginosa.
NPJ vaccines
Wang X,Liu C,Rcheulishvili N,Papukashvili D,Xie F,Zhao J,Hu X,Yu K,Yang N,Pan X,Liu X,Wang PG,He Y
Thu May 25 00:00:00 UTC 2023
Antioxidant hepatic lipid metabolism can be promoted by orally administered inorganic nanoparticles.
Nature communications
Cai J,Peng J,Feng J,Li R,Ren P,Zang X,Wu Z,Lu Y,Luo L,Hu Z,Wang J,Dai X,Zhao P,Wang J,Yan M,Liu J,Deng R,Wang D
Tue Jun 20 00:00:00 UTC 2023
A genome-wide CRISPR-Cas9 knockout screen identifies FSP1 as the warfarin-resistant vitamin K reductase.
Nature communications
Jin DY,Chen X,Liu Y,Williams CM,Pedersen LC,Stafford DW,Tie JK
Tue Feb 14 00:00:00 UTC 2023
A single-chain fragment constant design enables easy production of a monovalent blood-brain barrier transporter and provides an improved brain uptake at elevated doses.
Journal of neurochemistry
Morrison JI,Metzendorf NG,Rofo F,Petrovic A,Hultqvist G
Mon May 01 00:00:00 UTC 2023
Proximity labeling reveals OTUD3 as a DNA-binding deubiquitinase of cGAS.
Cell reports
Lyu H,Sun L,Guan Z,Li J,Yin C,Zhang Y,Jiang H
Tue Apr 25 00:00:00 UTC 2023
A human antibody potently neutralizes RSV by targeting the conserved hydrophobic region of prefusion F.
Science China. Life sciences
Yi C,Su C,Sun X,Lu X,Si C,Liu C,Yang Z,Yuan H,Huang Y,Wen J,He Y,Zhang Y,Ma L,Cong Y,Zhao G,Ling Z,Wang B,Sun B
Sat Apr 01 00:00:00 UTC 2023
Metabolic enzyme UAP1 mediates IRF3 pyrophosphorylation to facilitate innate immune response.
Molecular cell
Yang S,Jin S,Xian H,Zhao Z,Wang L,Wu Y,Zhou L,Li M,Cui J
Thu Jan 19 00:00:00 UTC 2023
SUE4, a novel PIN1-interacting membrane protein, regulates acropetal auxin transport in response to sulfur deficiency.
The New phytologist
Zhao Q,Zhao PX,Wu Y,Zhong CQ,Liao H,Li CY,Fu XD,Fang P,Xu P,Xiang CB
Sun Jan 01 00:00:00 UTC 2023
Genetically encoded chemical crosslinking of RNA in vivo.
Nature chemistry
Sun W,Wang N,Liu H,Yu B,Jin L,Ren X,Shen Y,Wang L
Sun Jan 01 00:00:00 UTC 2023
EgSeverin and Eg14-3-3zeta from Echinococcus granulosus are potential antigens for serological diagnosis of echinococcosis in dogs and sheep.
Microbial pathogenesis
Zhang X,Wei C,Lv Y,Mi R,Guo B,Rahman SU,Zhang Y,Cheng L,Jia H,Huang Y,Han X,Gong H,Chen Z
Thu Jun 01 00:00:00 UTC 2023
Development and preclinical evaluation of virus-like particle vaccine against COVID-19 infection.
Allergy
Yilmaz IC,Ipekoglu EM,Bulbul A,Turay N,Yildirim M,Evcili I,Yilmaz NS,Guvencli N,Aydin Y,Gungor B,Saraydar B,Bartan AG,Ibibik B,Bildik T,Baydemir İ,Sanli HA,Kayaoglu B,Ceylan Y,Yildirim T,Abras I,Ayanoglu IC,Cam SB,Ciftci Dede E,Gizer M,Erganis O,Sarac F,Uzar S,Enul H,Adiay C,Aykut G,Polat H,Yildirim IS,Tekin S,Korukluoglu G,Zeytin HE,Korkusuz P,Gursel I,Gursel M
Sat Jan 01 00:00:00 UTC 2022
HK1 from hepatic stellate cell-derived extracellular vesicles promotes progression of hepatocellular carcinoma.
Nature metabolism
Chen QT,Zhang ZY,Huang QL,Chen HZ,Hong WB,Lin T,Zhao WX,Wang XM,Ju CY,Wu LZ,Huang YY,Hou PP,Wang WJ,Zhou D,Deng X,Wu Q
Sat Oct 01 00:00:00 UTC 2022
Precise location of two novel linear epitopes on the receptor-binding domain surface of MERS-CoV spike protein recognized by two different monoclonal antibodies.
International journal of biological macromolecules
Wang P,Ding P,Wei Q,Liu H,Liu Y,Li Q,Xing Y,Li G,Zhou E,Zhang G
Sat Jan 15 00:00:00 UTC 2022
A Potent Neutralizing Nanobody Targeting the Spike Receptor-Binding Domain of SARS-CoV-2 and the Structural Basis of Its Intimate Binding.
Frontiers in immunology
Yang J,Lin S,Sun H,Chen Z,Yang F,Lin X,Guo L,Wang L,Wen A,Zhang X,Dai Y,He B,Cao Y,Dong H,Liu X,Chen B,Li J,Zhao Q,Lu G
Wed Jun 08 00:00:00 UTC 2022
Development of DNA Vaccine Candidate against SARS-CoV-2.
Viruses
Wang X,Rcheulishvili N,Cai J,Liu C,Xie F,Hu X,Yang N,Hou M,Papukashvili D,He Y,Wang PG
Sun May 15 00:00:00 UTC 2022
Transferrin Receptor Binding BBB-Shuttle Facilitates Brain Delivery of Anti-Aβ-Affibodies.
Pharmaceutical research
Faresjö R,Lindberg H,Ståhl S,Löfblom J,Syvänen S,Sehlin D
Fri Jul 01 00:00:00 UTC 2022
The Cycas genome and the early evolution of seed plants.
Nature plants
Liu Y,Wang S,Li L,Yang T,Dong S,Wei T,Wu S,Liu Y,Gong Y,Feng X,Ma J,Chang G,Huang J,Yang Y,Wang H,Liu M,Xu Y,Liang H,Yu J,Cai Y,Zhang Z,Fan Y,Mu W,Sahu SK,Liu S,Lang X,Yang L,Li N,Habib S,Yang Y,Lindstrom AJ,Liang P,Goffinet B,Zaman S,Wegrzyn JL,Li D,Liu J,Cui J,Sonnenschein EC,Wang X,Ruan J,Xue JY,Shao ZQ,Song C,Fan G,Li Z,Zhang L,Liu J,Liu ZJ,Jiao Y,Wang XQ,Wu H,Wang E,Lisby M,Yang H,Wang J,Liu X,Xu X,Li N,Soltis PS,Van de Peer Y,Soltis DE,Gong X,Liu H,Zhang S
Fri Apr 01 00:00:00 UTC 2022
Synonymous Codons and Hydrophobicity Optimization of Post-translational Signal Peptide PelB Increase Phage Display Efficiency of DARPins.
ACS synthetic biology
Kulmala A,Lappalainen M,Lamminmäki U,Huovinen T
Fri Oct 21 00:00:00 UTC 2022
Bispecific Antibody Expressed by an Oncolytic Herpes Simplex Virus Type 2 Can Transform Heterologous T Cells Into Uniform Tumor Killer Cells.
Human gene therapy
Jin J,Wang R,Yang J,Hu H,Wang D,Cai L,Fang Z,Dong S,Hu S,Wang Y,Liu B
Wed Jun 01 00:00:00 UTC 2022
Acetylation of fructose-bisphosphate aldolase-mediated glycolysis is essential for Bombyx mori nucleopolyhedrovirus infection.
Microbial pathogenesis
Gao X,Xu S,Mo Y,Zhu Y,Chen X,Miao M,Quan Y,Yu W
Thu Sep 01 00:00:00 UTC 2022
Acetylation of fructose-bisphosphate aldolase-mediated glycolysis is essential for Bombyx mori nucleopolyhedrovirus infection.
Microbial pathogenesis
Gao X,Xu S,Mo Y,Zhu Y,Chen X,Miao M,Quan Y,Yu W
Thu Sep 01 00:00:00 UTC 2022
Unique binding pattern for a lineage of human antibodies with broad reactivity against influenza A virus.
Nature communications
Sun X,Liu C,Lu X,Ling Z,Yi C,Zhang Z,Li Z,Jin M,Wang W,Tang S,Wang F,Wang F,Wangmo S,Chen S,Li L,Ma L,Zhang Y,Yang Z,Dong X,Qian Z,Ding J,Wang D,Cong Y,Sun B
Mon May 02 00:00:00 UTC 2022
PEBP balances apoptosis and autophagy in whitefly upon arbovirus infection.
Nature communications
Wang S,Guo H,Zhu-Salzman K,Ge F,Sun Y
Fri Feb 11 00:00:00 UTC 2022
A potent neutralizing and protective antibody against a conserved continuous epitope on HSV glycoprotein D.
Antiviral research
Tian R,Ju F,Yu M,Liang Z,Xu Z,Zhao M,Qin Y,Lin Y,Huang X,Chang Y,Li S,Ren W,Lin C,Xia N,Huang C
Sun May 01 00:00:00 UTC 2022
Neutralization Epitopes in Trimer and Pentamer Complexes Recognized by Potent Cytomegalovirus-Neutralizing Human Monoclonal Antibodies.
Microbiology spectrum
Ai Y,Wu C,Zhang M,Jaijyan DK,Liu T,Zan L,Li N,Yu W,Wang Y,Yuan X,Li C,Zheng W,Zhu H,Liao HX
Wed Dec 21 00:00:00 UTC 2022
A Novel Human Neutralizing mAb Recognizes Delta, Gamma and Omicron Variants of SARS-CoV-2 and Can Be Used in Combination with Sotrovimab.
International journal of molecular sciences
Passariello M,Ferrucci V,Sasso E,Manna L,Lembo RR,Pascarella S,Fusco G,Zambrano N,Zollo M,De Lorenzo C
Mon May 16 00:00:00 UTC 2022
Anti-TGF-β/PD-L1 bispecific antibody promotes T cell infiltration and exhibits enhanced antitumor activity in triple-negative breast cancer.
Journal for immunotherapy of cancer
Yi M,Wu Y,Niu M,Zhu S,Zhang J,Yan Y,Zhou P,Dai Z,Wu K
Thu Dec 01 00:00:00 UTC 2022
A lncRNA fine-tunes salicylic acid biosynthesis to balance plant immunity and growth.
Cell host & microbe
Liu N,Xu Y,Li Q,Cao Y,Yang D,Liu S,Wang X,Mi Y,Liu Y,Ding C,Liu Y,Li Y,Yuan YW,Gao G,Chen J,Qian W,Zhang X
Wed Aug 10 00:00:00 UTC 2022
Apostichopus japonicus matrix metalloproteinase-16 might act as a pattern recognition receptor.
Fish & shellfish immunology
Jiang J,Gao S,Chen Z,Guan X,Zhang F,Li L,Zhao Z,Zhao L,Xiao Y,Dong Y,Zhou Z
Tue Feb 01 00:00:00 UTC 2022
A Secreted Form of the Hepatitis E Virus ORF2 Protein: Design Strategy, Antigenicity and Immunogenicity.
Viruses
Chen Z,Guo S,Li G,Ying D,Wen G,Fang M,Wang Y,Tang Z,Zheng Z,Xia N
Mon Sep 26 00:00:00 UTC 2022
Recombinant Antibody-Based and Computer-Aided Comprehensive Analysis of Antibody's Equivalent Recognition Mechanism of Alternariol and Alternariol Monomethyl Ether.
Frontiers in chemistry
Wang Z,Chen L,Zheng P,Wang J,Ren Z,Zhang H,Zhang L,Jiang H
Sat Jul 16 00:00:00 UTC 2022
Identification of a Novel Neutralizing Epitope on the N-Terminal Domain of the Human Coronavirus 229E Spike Protein.
Journal of virology
Shi J,Shi Y,Xiu R,Wang G,Liang R,Jiao Y,Shen Z,Zhu C,Peng G
Wed Feb 23 00:00:00 UTC 2022
Modular assembly of the principal microtubule nucleator γ-TuRC.
Nature communications
Würtz M,Zupa E,Atorino ES,Neuner A,Böhler A,Rahadian AS,Vermeulen BJA,Tonon G,Eustermann S,Schiebel E,Pfeffer S
Tue Jan 25 00:00:00 UTC 2022
Characterization of a novel thermophilic metagenomic GH5 endoglucanase heterologously expressed in Escherichia coli and Saccharomyces cerevisiae.
Biotechnology for biofuels and bioproducts
Escuder-Rodríguez JJ,González-Suarez M,deCastro ME,Saavedra-Bouza A,Becerra M,González-Siso MI
Thu Jul 07 00:00:00 UTC 2022
TMUB1 is an endoplasmic reticulum-resident escortase that promotes the p97-mediated extraction of membrane proteins for degradation.
Molecular cell
Wang L,Li J,Wang Q,Ge MX,Ji J,Liu D,Wang Z,Cao Y,Zhang Y,Zhang ZR
Thu Sep 15 00:00:00 UTC 2022
7,8-Dihydroxyflavone ameliorates mitochondrial impairment and motor dysfunction in the α-synuclein 1-103 transgenic mice.
Neurobiology of disease
Tian Y,Pan L,Yuan X,Xiong M,Zhang Z,Meng L,Zheng Y,Bu L,Xu X,Zhang Z
Fri Jul 01 00:00:00 UTC 2022
Novel Bi-Specific Immuno-Modulatory Tribodies Potentiate T Cell Activation and Increase Anti-Tumor Efficacy.
International journal of molecular sciences
Passariello M,Yoshioka A,Takahashi K,Hashimoto SI,Rapuano Lembo R,Manna L,Nakamura K,De Lorenzo C
Wed Mar 23 00:00:00 UTC 2022
Establishment of a carbohydrate binding module-based lateral flow immunoassay method for identifying hyaluronic acid.
International journal of biological macromolecules
Mei X,Sun M,Zhang Y,Shen J,Li J,Xue C,Chang Y
Sat Dec 31 00:00:00 UTC 2022
Possible Cross-Reactivity of Feline and White-Tailed Deer Antibodies against the SARS-CoV-2 Receptor Binding Domain.
Journal of virology
Hancock TJ,Hickman P,Kazerooni N,Kennedy M,Kania SA,Dennis M,Szafranski N,Gerhold R,Su C,Masi T,Smith S,Sparer TE
Wed Apr 27 00:00:00 UTC 2022
A Five-Helix-Based SARS-CoV-2 Fusion Inhibitor Targeting Heptad Repeat 2 Domain against SARS-CoV-2 and Its Variants of Concern.
Viruses
Xing L,Xu X,Xu W,Liu Z,Shen X,Zhou J,Xu L,Pu J,Yang C,Huang Y,Lu L,Jiang S,Liu S
Sun Mar 13 00:00:00 UTC 2022
TurboID Screening of the OmpP2 Protein Reveals Host Proteins Involved in Recognition and Phagocytosis of Glaesserella parasuis by iPAM Cells.
Microbiology spectrum
Jiang C,Ma N,Cao H,Zeng W,Ren J,Hu Y,Zhou J,Zhang M,Li C,Lang Y,Li W,He Q
Wed Oct 26 00:00:00 UTC 2022
TurboID Screening of the OmpP2 Protein Reveals Host Proteins Involved in Recognition and Phagocytosis of Glaesserella parasuis by iPAM Cells.
Microbiology spectrum
Jiang C,Ma N,Cao H,Zeng W,Ren J,Hu Y,Zhou J,Zhang M,Li C,Lang Y,Li W,He Q
Wed Oct 26 00:00:00 UTC 2022
Tankyrase-1-mediated degradation of Golgin45 regulates glycosyltransferase trafficking and protein glycosylation in Rab2-GTP-dependent manner.
Communications biology
Yue X,Tiwari N,Zhu L,Ngo HDT,Lim JM,Gim B,Jing S,Wang Y,Qian Y,Lee I
Tue Dec 07 00:00:00 UTC 2021
Expanding virus susceptibility spectrum of MDBK cells by expressing host receptors nectin 4 and TfR.
Journal of virological methods
Han P,Zhang X,Yang S,Dai X,Lv Q,Hua D
Mon Mar 01 00:00:00 UTC 2021
Prokaryotic expression of chimeric GFP-hFc protein as a potential immune-based tool.
Molecular biology research communications
Nguyen TT,Vo-Nguyen HV,Tran-Van H
Wed Sep 01 00:00:00 UTC 2021
Deacetylation of BmAda3 is required for cell apoptosis caused by Bombyx mori nucleopolyhedrovirus infection.
Archives of insect biochemistry and physiology
Zhu Y,Hu M,Ngowo J,Gao X,Chen X,Yan H,Yu W
Fri Oct 01 00:00:00 UTC 2021
A Novel Bifunctional Wax Ester Synthase Involved in Early Triacylglycerol Accumulation in Unicellular Green Microalga Haematococcus pluvialis Under High Light Stress.
Frontiers in bioengineering and biotechnology
Ma H,Zheng J,Li Y,Zhao L,Zou S,Hu Q,Han D
Fri Feb 04 00:00:00 UTC 2022
A bispecific nanobody targeting the dimerization interface of epidermal growth factor receptor: Evidence for tumor suppressive actions in vitro and in vivo.
Biochemical and biophysical research communications
Xu Z,Qiu C,Wen B,Wang S,Zhu L,Zhao L,Li H
Fri Apr 09 00:00:00 UTC 2021
IRF3-binding lncRNA-ISIR strengthens interferon production in viral infection and autoinflammation.
Cell reports
Xu J,Wang P,Li Z,Li Z,Han D,Wen M,Zhao Q,Zhang L,Ma Y,Liu W,Jiang M,Zhang X,Cao X
Tue Nov 02 00:00:00 UTC 2021
Anti-osteosarcoma effect of antiserum against cross antigen TPD52 between osteosarcoma and Trichinella spiralis.
Parasites & vectors
Yue TT,Zhang N,Li JH,Lu XY,Wang XC,Li X,Zhang HB,Cheng SQ,Wang BB,Gong PT,Zhang XC
Sun Sep 26 00:00:00 UTC 2021
Recombinant Human SCARB2 Expressed in Escherichia coli and its Potential in Enterovirus 71 Blockage.
Iranian journal of science and technology. Transaction A, Science
Vo-Nguyen HV,Nguyen TT,Vu HT,Nguyen TT,Hoang QC,Tran TL,Tran-Van H
Thu Jul 07 00:00:00 UTC 2022
Development of nanobody-based flow-through dot ELISA and lateral-flow immunoassay for rapid detection of 3-phenoxybenzoic acid.
Analytical methods : advancing methods and applications
Zhang C,Wu X,Li D,Hu J,Wan D,Zhang Z,Hammock BD
Wed Apr 14 00:00:00 UTC 2021
The oncomicropeptide APPLE promotes hematopoietic malignancy by enhancing translation initiation.
Molecular cell
Sun L,Wang W,Han C,Huang W,Sun Y,Fang K,Zeng Z,Yang Q,Pan Q,Chen T,Luo X,Chen Y
Thu Nov 04 00:00:00 UTC 2021
Expression and Purification of Yeast-derived GPCR, Gα and Gβγ Subunits for Structural and Dynamic Studies.
Bio-protocol
Zhao W,Wang X,Ye L
Sat Feb 20 00:00:00 UTC 2021
Epigallocatechin gallate from green tea effectively blocks infection of SARS-CoV-2 and new variants by inhibiting spike binding to ACE2 receptor.
Cell & bioscience
Liu J,Bodnar BH,Meng F,Khan AI,Wang X,Saribas S,Wang T,Lohani SC,Wang P,Wei Z,Luo J,Zhou L,Wu J,Luo G,Li Q,Hu W,Ho W
Mon Aug 30 00:00:00 UTC 2021
Bacterial expression of a snake venom metalloproteinase inhibitory protein from the North American opossum (D.virginiana).
Toxicon : official journal of the International Society on Toxinology
Werner RM,Miling LM,Elliott BM,Hawes MR,Wickens JM,Webber DE
Fri Apr 30 00:00:00 UTC 2021
Comprehensive mapping of binding hot spots of SARS-CoV-2 RBD-specific neutralizing antibodies for tracking immune escape variants.
Genome medicine
Yi C,Sun X,Lin Y,Gu C,Ding L,Lu X,Yang Z,Zhang Y,Ma L,Gu W,Qu A,Zhou X,Li X,Xu J,Ling Z,Xie Y,Lu H,Sun B
Thu Oct 14 00:00:00 UTC 2021
Glutaredoxins regulate maize inflorescence meristem development via redox control of TGA transcriptional activity.
Nature plants
Yang RS,Xu F,Wang YM,Zhong WS,Dong L,Shi YN,Tang TJ,Sheng HJ,Jackson D,Yang F
Wed Dec 01 00:00:00 UTC 2021
Structural Insights into JAK2 Inhibition by Ruxolitinib, Fedratinib, and Derivatives Thereof.
Journal of medicinal chemistry
Davis RR,Li B,Yun SY,Chan A,Nareddy P,Gunawan S,Ayaz M,Lawrence HR,Reuther GW,Lawrence NJ,Schönbrunn E
Thu Feb 25 00:00:00 UTC 2021
The SESAME complex regulates cell senescence through the generation of acetyl-CoA.
Nature metabolism
Chen W,Yu X,Wu Y,Tang J,Yu Q,Lv X,Zha Z,Hu B,Li X,Chen J,Ma L,Workman JL,Li S
Thu Jul 01 00:00:00 UTC 2021
A bivalent protein targeting glycans and HR1 domain in spike protein potently inhibited infection of SARS-CoV-2 and other human coronaviruses.
Cell & bioscience
Cai Y,Xu W,Tang J,Cao N,Lan Q,Lu L,Jiang S
Thu Jul 08 00:00:00 UTC 2021
EAT-18 is an essential auxiliary protein interacting with the non-alpha nAChR subunit EAT-2 to form a functional receptor.
PLoS pathogens
Choudhary S,Buxton SK,Puttachary S,Verma S,Mair GR,McCoy CJ,Reaves BJ,Wolstenholme AJ,Martin RJ,Robertson AP
Wed Apr 01 00:00:00 UTC 2020
Interactions of ferritin with scavenger receptor class A members.
The Journal of biological chemistry
Yu B,Cheng C,Wu Y,Guo L,Kong D,Zhang Z,Wang Y,Zheng E,Liu Y,He Y
Fri Nov 13 00:00:00 UTC 2020
Multiple functions of thymosin β4 in the pearl oyster Pinctada fucata suggest its multiple potential roles in artificial pearl culture.
Fish & shellfish immunology
Bai L,He W,Fan S,Liu B,Zhou T,Zhang D,Zhang D,Yu D
Sat Aug 01 00:00:00 UTC 2020
Decreased Expression of the Host Long-Noncoding RNA-GM Facilitates Viral Escape by Inhibiting the Kinase activity TBK1 via S-glutathionylation.
Immunity
Wang Y,Wang P,Zhang Y,Xu J,Li Z,Li Z,Zhou Z,Liu L,Cao X
Tue Dec 15 00:00:00 UTC 2020
Spike Protein of SARS-CoV-2 Activates Macrophages and Contributes to Induction of Acute Lung Inflammations in Mice.
bioRxiv : the preprint server for biology
Cao X,Tian Y,Nguyen V,Zhang Y,Gao C,Yin R,Carver W,Fan D,Albrecht H,Cui T,Tan W
Mon Dec 07 00:00:00 UTC 2020
Detection of SARS-CoV-2-Specific Humoral and Cellular Immunity in COVID-19 Convalescent Individuals.
Immunity
Ni L,Ye F,Cheng ML,Feng Y,Deng YQ,Zhao H,Wei P,Ge J,Gou M,Li X,Sun L,Cao T,Wang P,Zhou C,Zhang R,Liang P,Guo H,Wang X,Qin CF,Chen F,Dong C
Tue Jun 16 00:00:00 UTC 2020
PII Signal Transduction Protein GlnK Alleviates Feedback Inhibition of N-Acetyl-l-Glutamate Kinase by l-Arginine in Corynebacterium glutamicum.
Applied and environmental microbiology
Xu M,Tang M,Chen J,Yang T,Zhang X,Shao M,Xu Z,Rao Z
Wed Apr 01 00:00:00 UTC 2020
Sorting nexin 17 (SNX17) links endosomal sorting to Eps15 homology domain protein 1 (EHD1)-mediated fission machinery.
The Journal of biological chemistry
Dhawan K,Naslavsky N,Caplan S
Fri Mar 20 00:00:00 UTC 2020
Developing Covalent Protein Drugs via Proximity-Enabled Reactive Therapeutics.
Cell
Li Q,Chen Q,Klauser PC,Li M,Zheng F,Wang N,Li X,Zhang Q,Fu X,Wang Q,Xu Y,Wang L
Thu Jul 09 00:00:00 UTC 2020
Expression and characterization of a novel alginate-binding protein: A promising tool for investigating alginate.
Carbohydrate polymers
Mei X,Chang Y,Shen J,Zhang Y,Xue C
Thu Oct 15 00:00:00 UTC 2020
Constructing a human complex type N-linked glycosylation pathway in Kluyveromyces marxianus.
PloS one
Lee MH,Hsu TL,Lin JJ,Lin YJ,Kao YY,Chang JJ,Li WH
Mon Aug 31 00:00:00 UTC 2020
Conditioned Medium from Cells Overexpressing TDP-43 Alters the Metabolome of Recipient Cells.
Cells
Hergesheimer R,Lanznaster D,Bourgeais J,Hérault O,Vourc'h P,Andres CR,Corcia P,Blasco H
Tue Sep 29 00:00:00 UTC 2020
An albumin‑binding domain and targeting peptide‑based recombinant protein and its enediyne‑integrated analogue exhibit directional delivery and potent inhibitory activity on pancreatic cancer with K‑ras mutation.
Oncology reports
Sheng W,Geng J,Li L,Shang Y,Jiang M,Zhen Y
Sun Mar 01 00:00:00 UTC 2020
Human-IgG-Neutralizing Monoclonal Antibodies Block the SARS-CoV-2 Infection.
Cell reports
Wan J,Xing S,Ding L,Wang Y,Gu C,Wu Y,Rong B,Li C,Wang S,Chen K,He C,Zhu D,Yuan S,Qiu C,Zhao C,Nie L,Gao Z,Jiao J,Zhang X,Wang X,Ying T,Wang H,Xie Y,Lu Y,Xu J,Lan F
Tue Jul 21 00:00:00 UTC 2020
Cryo-EM structure of the mitochondrial protein-import channel TOM complex at near-atomic resolution.
Nature structural & molecular biology
Tucker K,Park E
Sun Dec 01 00:00:00 UTC 2019
STING signaling remodels the tumor microenvironment by antagonizing myeloid-derived suppressor cell expansion.
Cell death and differentiation
Zhang CX,Ye SB,Ni JJ,Cai TT,Liu YN,Huang DJ,Mai HQ,Chen QY,He J,Zhang XS,Zeng YX,Li J,Cui J
Fri Nov 01 00:00:00 UTC 2019
A novel crosslinking protocol stabilizes amyloid β oligomers capable of inducing Alzheimer's-associated pathologies.
Journal of neurochemistry
Cline EN,Das A,Bicca MA,Mohammad SN,Schachner LF,Kamel JM,DiNunno N,Weng A,Paschall JD,Bu RL,Khan FM,Rollins MG,Ives AN,Shekhawat G,Nunes-Tavares N,de Mello FG,Compton PD,Kelleher NL,Klein WL
Fri Mar 01 00:00:00 UTC 2019
A novel crosslinking protocol stabilizes amyloid β oligomers capable of inducing Alzheimer's-associated pathologies.
Journal of neurochemistry
Cline EN,Das A,Bicca MA,Mohammad SN,Schachner LF,Kamel JM,DiNunno N,Weng A,Paschall JD,Bu RL,Khan FM,Rollins MG,Ives AN,Shekhawat G,Nunes-Tavares N,de Mello FG,Compton PD,Kelleher NL,Klein WL
Fri Mar 01 00:00:00 UTC 2019
The retroviral accessory proteins S2, Nef, and glycoMA use similar mechanisms for antagonizing the host restriction factor SERINC5.
The Journal of biological chemistry
Ahmad I,Li S,Li R,Chai Q,Zhang L,Wang B,Yu C,Zheng YH
Fri Apr 26 00:00:00 UTC 2019
Novel Mussel-Inspired Universal Surface Functionalization Strategy: Protein-Based Coating with Residue-Specific Post-Translational Modification in Vivo.
ACS applied materials & interfaces
Qi H,Zheng W,Zhang C,Zhou X,Zhang L
Wed Apr 03 00:00:00 UTC 2019
Receptor-Mediated Field Effect Transistor Biosensor for Real-Time Monitoring of Glutamate Release from Primary Hippocampal Neurons.
Analytical chemistry
Li YT,Jin X,Tang L,Lv WL,Xiao MM,Zhang ZY,Gao C,Zhang GJ
Tue Jul 02 00:00:00 UTC 2019
Characterization of PPIB interaction in the P3H1 ternary complex and implications for its pathological mutations.
Cellular and molecular life sciences : CMLS
Wu J,Zhang W,Xia L,Feng L,Shu Z,Zhang J,Ye W,Zeng N,Zhou A
Tue Oct 01 00:00:00 UTC 2019
Characterization of two engineered dimeric Zika virus envelope proteins as immunogens for neutralizing antibody selection and vaccine design.
The Journal of biological chemistry
Yang C,Zeng F,Gao X,Zhao S,Li X,Liu S,Li N,Deng C,Zhang B,Gong R
Fri Jul 05 00:00:00 UTC 2019
Vancomycin does not affect the enzymatic activities of purified VanSA.
PloS one
Upton EC,Maciunas LJ,Loll PJ
Wed Oct 16 00:00:00 UTC 2019
Identification of a cellularly active SIRT6 allosteric activator.
Nature chemical biology
Huang Z,Zhao J,Deng W,Chen Y,Shang J,Song K,Zhang L,Wang C,Lu S,Yang X,He B,Min J,Hu H,Tan M,Xu J,Zhang Q,Zhong J,Sun X,Mao Z,Lin H,Xiao M,Chin YE,Jiang H,Xu Y,Chen G,Zhang J
Sat Dec 01 00:00:00 UTC 2018
Identification of a cellularly active SIRT6 allosteric activator.
Nature chemical biology
Huang Z,Zhao J,Deng W,Chen Y,Shang J,Song K,Zhang L,Wang C,Lu S,Yang X,He B,Min J,Hu H,Tan M,Xu J,Zhang Q,Zhong J,Sun X,Mao Z,Lin H,Xiao M,Chin YE,Jiang H,Xu Y,Chen G,Zhang J
Sat Dec 01 00:00:00 UTC 2018
Sites and Functional Consequence of Alkylphenol Anesthetic Binding to Kv1.2 Channels.
Molecular neurobiology
Bu W,Liang Q,Zhi L,Maciunas L,Loll PJ,Eckenhoff RG,Covarrubias M
Thu Feb 01 00:00:00 UTC 2018
Sites and Functional Consequence of Alkylphenol Anesthetic Binding to Kv1.2 Channels.
Molecular neurobiology
Bu W,Liang Q,Zhi L,Maciunas L,Loll PJ,Eckenhoff RG,Covarrubias M
Thu Feb 01 00:00:00 UTC 2018
An alternative conformation of human TrpRS suggests a role of zinc in activating non-enzymatic function.
RNA biology
Xu X,Zhou H,Zhou Q,Hong F,Vo MN,Niu W,Wang Z,Xiong X,Nakamura K,Wakasugi K,Schimmel P,Yang XL
Fri Nov 02 00:00:00 UTC 2018
An alternative conformation of human TrpRS suggests a role of zinc in activating non-enzymatic function.
RNA biology
Xu X,Zhou H,Zhou Q,Hong F,Vo MN,Niu W,Wang Z,Xiong X,Nakamura K,Wakasugi K,Schimmel P,Yang XL
Fri Nov 02 00:00:00 UTC 2018
GlpR Is a Direct Transcriptional Repressor of Fructose Metabolic Genes in Haloferax volcanii.
Journal of bacteriology
Martin JH,Sherwood Rawls K,Chan JC,Hwang S,Martinez-Pastor M,McMillan LJ,Prunetti L,Schmid AK,Maupin-Furlow JA
Sat Sep 01 00:00:00 UTC 2018
GlpR Is a Direct Transcriptional Repressor of Fructose Metabolic Genes in Haloferax volcanii.
Journal of bacteriology
Martin JH,Sherwood Rawls K,Chan JC,Hwang S,Martinez-Pastor M,McMillan LJ,Prunetti L,Schmid AK,Maupin-Furlow JA
Sat Sep 01 00:00:00 UTC 2018
Interaction of Calmodulin with the cSH2 Domain of the p85 Regulatory Subunit.
Biochemistry
Wang G,Zhang M,Jang H,Lu S,Lin S,Chen G,Nussinov R,Zhang J,Gaponenko V
Tue Mar 27 00:00:00 UTC 2018
High-efficiency production of human serum albumin in the posterior silk glands of transgenic silkworms, Bombyx mori L.
PloS one
Qian Q,You Z,Ye L,Che J,Wang Y,Wang S,Zhong B
Mon Feb 26 00:00:00 UTC 2018
High-efficiency production of human serum albumin in the posterior silk glands of transgenic silkworms, Bombyx mori L.
PloS one
Qian Q,You Z,Ye L,Che J,Wang Y,Wang S,Zhong B
Mon Feb 26 00:00:00 UTC 2018
In vitro Analysis of Ubiquitin-like Protein Modification in Archaea.
Bio-protocol
Fu X,Adams Z,Maupin-Furlow JA
Sun May 20 00:00:00 UTC 2018
In vitro Analysis of Ubiquitin-like Protein Modification in Archaea.
Bio-protocol
Fu X,Adams Z,Maupin-Furlow JA
Sun May 20 00:00:00 UTC 2018
3-Hydroxyisobutyrate Dehydrogenase Is Involved in Both, Valine and Isoleucine Degradation in Arabidopsis thaliana.
Plant physiology
Schertl P,Danne L,Braun HP
Fri Sep 01 00:00:00 UTC 2017
3-Hydroxyisobutyrate Dehydrogenase Is Involved in Both, Valine and Isoleucine Degradation in Arabidopsis thaliana.
Plant physiology
Schertl P,Danne L,Braun HP
Fri Sep 01 00:00:00 UTC 2017
5-Aminolevulinic Acid Dehydratase Gene Dosage Affects Programmed Cell Death and Immunity.
Plant physiology
Chai Q,Shang X,Wu S,Zhu G,Cheng C,Cai C,Wang X,Guo W
Fri Sep 01 00:00:00 UTC 2017
5-Aminolevulinic Acid Dehydratase Gene Dosage Affects Programmed Cell Death and Immunity.
Plant physiology
Chai Q,Shang X,Wu S,Zhu G,Cheng C,Cai C,Wang X,Guo W
Fri Sep 01 00:00:00 UTC 2017
A human scFv antibody that targets and neutralizes high molecular weight pathogenic amyloid-β oligomers.
Journal of neurochemistry
Sebollela A,Cline EN,Popova I,Luo K,Sun X,Ahn J,Barcelos MA,Bezerra VN,Lyra E Silva NM,Patel J,Pinheiro NR,Qin LA,Kamel JM,Weng A,DiNunno N,Bebenek AM,Velasco PT,Viola KL,Lacor PN,Ferreira ST,Klein WL
Fri Sep 01 00:00:00 UTC 2017
A human scFv antibody that targets and neutralizes high molecular weight pathogenic amyloid-β oligomers.
Journal of neurochemistry
Sebollela A,Cline EN,Popova I,Luo K,Sun X,Ahn J,Barcelos MA,Bezerra VN,Lyra E Silva NM,Patel J,Pinheiro NR,Qin LA,Kamel JM,Weng A,DiNunno N,Bebenek AM,Velasco PT,Viola KL,Lacor PN,Ferreira ST,Klein WL
Fri Sep 01 00:00:00 UTC 2017
Effects of microvirin monomers and oligomers on hepatitis C virus.
Bioscience reports
Min YQ,Duan XC,Zhou YD,Kulinich A,Meng W,Cai ZP,Ma HY,Liu L,Zhang XL,Voglmeir J
Fri Jun 30 00:00:00 UTC 2017
Effects of microvirin monomers and oligomers on hepatitis C virus.
Bioscience reports
Min YQ,Duan XC,Zhou YD,Kulinich A,Meng W,Cai ZP,Ma HY,Liu L,Zhang XL,Voglmeir J
Fri Jun 30 00:00:00 UTC 2017
Construction of an expression vector that uses the aph promoter for protein expression in Corynebacterium glutamicum.
Plasmid
Zhang W,Zhao Z,Yang Y,Liu X,Bai Z
Wed Nov 01 00:00:00 UTC 2017
Construction of an expression vector that uses the aph promoter for protein expression in Corynebacterium glutamicum.
Plasmid
Zhang W,Zhao Z,Yang Y,Liu X,Bai Z
Wed Nov 01 00:00:00 UTC 2017
Identification of a Novel Inhibitor against Middle East Respiratory Syndrome Coronavirus.
Viruses
Sun Y,Zhang H,Shi J,Zhang Z,Gong R
Thu Sep 14 00:00:00 UTC 2017
Identification of a Novel Inhibitor against Middle East Respiratory Syndrome Coronavirus.
Viruses
Sun Y,Zhang H,Shi J,Zhang Z,Gong R
Thu Sep 14 00:00:00 UTC 2017