Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: QEFLLRVEPQ NPVLSAGGSL FVNCSTDCPS SEKIALETSL SKELVASGMG WAAFNLSNVT GNSRILCSVY CNGSQITGSS NITVYRLPER VELAPLPPWQ PVGQNFTLRC QVEDGSPRTS LTVVLLRWEE ELSRQPAVEE PAEVTATVLA SRDDHGAPFS CRTELDMQPQ GLGLFVNTSA PRQLRTFVLP VTPPRLVAPR FLEVETSWPV DCTLDGLFPA SEAQVYLALG DQMLNATVMN HGDTLTATAT ATARADQEGA REIVCNVTLG GERREARENL TVFSFLGPIV NLSEPTAHEG STVTVSCMAG ARVQVTLDGV PAAAPGQPAQ LQLNATESDD GRSFFCSATL EVDGEFLHRN SSVQLRVLYG PKIDRATCPQ HLKWKDKTRH VLQCQARGNP YPELRCLKEG SSREVPVGIP FFVNVTHNGT YQCQASSSRG KYTLVVVMDI EAGSSHFVPV FVAVLLTLGV VTIVLALMYV FREHQRSGSY HVREESTYLP LTSMQPTEAM GEEPSRAE
CD50 (intracellular adhesion molecule 3, ICAM-3) is a transmembrane glycoprotein expressed by leukocytes, that serves as a counter-receptor for the lymphocyte function-associated antigen (LFA)-1 integrin. Besides functioning as an adhesive molecule that mediates e.g. the contact between T cells and antigen presenting cells, ICAM-3 regulates affinity of LFA-1 for ICAM-1 and induces T cell activation and proliferation. ICAM-3 plays an essential role in the initiation of the immune response both on T cells and antigen presenting cells and interacts also with CD209 (dendritic cell-specific ICAM-3-grabbing nonintegrin, DC-SIGN), a C-type lectin of dendritic cells and macrophages; this process is involved in dialogue between dendritic cells and granulocytes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD50; CDw50; ICAM-3; ICAM-R; Intercellular adhesion molecule 3; intercellular adhesion molecule-3
Gene Aliases: CD50; CDW50; ICAM-R; ICAM3
UniProt ID: (Human) P32942
Entrez Gene ID: (Human) 3385
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support