Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • ID3 Antibodies

          OriGene

          ID3 Monoclonal Antibody (OTI15F4), TrueMAB™

          View all (38) ID3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite ID3 Monoclonal Antibody (OTI15F4), TrueMAB™

          • Antibody Testing Data (19)
          ID3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          ID3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 19

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ID3 Antibody (CF500716) in ICC/IF

          Anti-ID3 mouse monoclonal antibody (TA500716) Immunofluorescent staining of COS7 cells transiently transfected by pCMV6-ENTRY ID3(RC200583). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ID3 Antibody in Immunocytochemistry (ICC/IF)
          ID3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ID3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ID3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ID3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ID3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ID3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ID3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ID3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ID3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ID3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ID3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ID3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ID3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ID3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ID3 Antibody in Western Blot (WB)
          ID3 Antibody in Western Blot (WB)
          ID3 Antibody in Flow Cytometry (Flow)
          ID3 Antibody in Flow Cytometry (Flow)

          Product Details

          CF500716

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:200
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:400
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:400
          -

          Flow Cytometry (Flow)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          OTI15F4

          Immunogen

          Full length human recombinant protein of human ID3 produced in HEK293T cell.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          1 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 8% trehalose

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)

          Target Information

          Members of the Id family of basic helix-loop-helix (bHLH) proteins include ID1, ID2, ID3 and ID4. They are ubiquitously expressed and dimerize with members of the class A and B HLH proteins. Due to the absence of the basic region, the resulting heterodimers cannot bind DNA. The ID-type proteins thus appear to negatively regulate DNA binding of bHLH proteins. Since ID1 inhibits DNA binding of E12 and Myo D, it apparently functions to inhibit muscle-specific gene expression. Under conditions that facilitate muscle cell differentiation, the ID protein levels fall, allowing E12 and/or E47 to form heterodimers with Myo D and myogenin, which in turn activate myogenic differentiation. It has been shown that expression of each of the ID proteins is strongly dependent on growth factor activation and that reduction of ID mRNA levels by antisense oligonucleotides leads to a delayed reentry of arrested cells into the cell cycle following growth factor stimulation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: bHLHb25; Class B basic helix-loop-helix protein 25; DNA-binding protein inhibitor ID-3; HEIR 1; Helix-loop-helix protein HEIR-1; HLH642; ID 3; ID-like protein inhibitor HLH 1R21; Inhibitor of differentiation 3; Inhibitor of DNA binding 3; inhibitor of DNA binding 3, dominant negative helix-loop-helix protein

          View more View less

          Gene Aliases: 1R21; BHLHB25; HEIR-1; HEIR1; ID3

          View more View less

          UniProt ID: (Human) Q02535

          View more View less

          Entrez Gene ID: (Human) 3399

          View more View less

          Function(s)
          transcription factor activity, sequence-specific DNA binding transcription corepressor activity protein binding transcription factor binding protein domain specific binding protein dimerization activity leptomycin B binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter metanephros development regulation of DNA replication transcription, DNA-templated multicellular organism development central nervous system development heart development muscle organ development circadian rhythm response to wounding neuron differentiation epithelial cell differentiation notochord development odontogenesis positive regulation of apoptotic process negative regulation of sequence-specific DNA binding transcription factor activity negative regulation of myoblast differentiation negative regulation of osteoblast differentiation negative regulation of transcription, DNA-templated regulation of cell cycle cellular response to leptomycin B
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-2w6sl:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0