Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Sequence of this protein is as follows: MSTNGDDHQV KDSLEQLRCH FTWELSIDDD EMPDLENRVL DQIEFLDTKY SVGIHNLLAY VKHLKGQNEE ALKSLKEAEN LMQEEHDNQA NVRSLVTWGN FAWMYYHMGR LAEAQTYLDK VENICKKLSN PFRYRMECPE IDCEEGWALL KCGGKNYERA KACFEKVLEV DPENPESSAG YAISAYRLDG FKLATKNHKP FSLLPLRQAV RLNPDNGYIK VLLALKLQDE GQEAEGEKYI EEALANMSSQ TYVFRYAAKF YRRKGSVDKA LELLKKALQE TPTSVLLHHQ IGLCYKAQMI QIKEATKGQP RGQNREKLDK MIRSAIFHFE SAVEKKPTFE VAHLDLARMY IEAGNHRKAE ENFQKLLCMK PVVEETMQDI HFHYGRFQEF QKKSDVNAII HYLKAIKIEQ ASLTRDKSIN SLKKLVLRKL RRKALDLESL SLLGFVYKLE GNMNEALEYY ERALRLAADF ENSVRQGP
Interferon-induced antiviral RNA-binding protein that specifically binds single-stranded RNA bearing a 5'-triphosphate group (PPP-RNA), thereby acting as a sensor of viral single-stranded RNAs and inhibiting expression of viral messenger RNAs. Single-stranded PPP-RNAs, which lack 2'-O-methylation of the 5' cap and bear a 5'-triphosphate group instead, are specific from viruses, providing a molecular signature to distinguish between self and non-self mRNAs by the host during viral infection. Directly binds PPP-RNA in a non-sequence-specific manner. Viruses evolved several ways to evade this restriction system such as encoding their own 2'-O-methylase for their mRNAs or by stealing host cap containing the 2'-O-methylation (cap snatching mechanism). Exhibits antiviral activity against several viruses including human papilloma and hepatitis C viruses.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IFI-56K; IFIT-1; interferon, alpha-inducible protein (MW 56kD); Interferon-induced 56 kDa protein; Interferon-induced protein with tetratricopeptide repeats 1; interferon-inducible mRNA 561
Gene Aliases: C56; G10P1; IFI-56; IFI-56K; IFI56; IFIT-1; IFIT1; IFNAI1; ISG56; P56; RNM561
UniProt ID: (Human) P09914
Entrez Gene ID: (Human) 3434
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support