Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • HIPPI Antibodies

          Proteintech

          IFT57 Polyclonal Antibody

          26 References
          View all (22) HIPPI antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite IFT57 Polyclonal Antibody

          • Antibody Testing Data (15)
          IFT57 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          IFT57 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 15

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          IFT57 Antibody (11083-1-AP) in ICC/IF

          Immunofluorescent analysis of MDCK cells using 11083-1-AP (IFT57 antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          IFT57 Antibody in Immunocytochemistry (ICC/IF)
          IFT57 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IFT57 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IFT57 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IFT57 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IFT57 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IFT57 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IFT57 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IFT57 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IFT57 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IFT57 Antibody in Western Blot (WB)
          IFT57 Antibody in Western Blot (WB)
          IFT57 Antibody in Western Blot (WB)
          IFT57 Antibody in Western Blot (WB)
          IFT57 Antibody in Immunoprecipitation (IP)

          Product Details

          11083-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          View 13 publications 13 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          View 10 publications 10 publications

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 2 publications 2 publications

          Not applicable (N/A)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Dog, Human, Mouse, Rat

          Published species

          Chicken, Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          IFT57 Fusion Protein Ag1566 (1-429 aa encoded by BC011899)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.35 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MTAALAVVT TSGLEDGVPR SRGEGTGEVV LERGPGAAYH MFVVMEDLVE KLKLLRYEEE FLRKSNLKAP SRHYFALPTN PGEQFYMFCT LAAWLINKAG RPFEQPQEYD DPNATISNIL SELRSFGRTA DFPPSKLKSG YGEHVCYVLD CFAEEALKYI GFTWKRPIYP VEELEEESVA EDDAELTLNK VDEEFVEEET DNEENFIDLN VLKAQTYHLD MNETAKQEDI LESTTDAAEW SLEVERVLPQ LKVTIRTDNK DWRIHVDQMH QHRSGIESAL KETKGFLDKL HNEITRTLEK ISSREKYINN QLENLVQEYR AAQAQLSEAK ERYQQGNGGV TERTRLLSEV MEELEKVKQE MEEKGSSMTD GAPLVKIKQS LTKLKQETVE MDIRIGIVEH TLLQSKLKEK SNMTRNMHAT VIPEPATGFY (1-429 aa encoded by BC011899)

          Target Information

          HIPPI is required for the formation of cilia. Plays an indirect role in sonic hedgehog signaling, cilia being required for all activity of the hedgehog pathway. Has pro-apoptotic function via its interaction with HIP1, leading to recruit caspase-8 (CASP8) and trigger apoptosis. Has the ability to bind DNA sequence motif 5'-AAAGACATG-3' present in the promoter of caspase genes such as CASP1, CASP8 and CASP10, suggesting that it may act as a transcription regulator; however the relevance of such function remains unclear.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Dermal papilla-derived protein 8; estrogen-related receptor beta like 1; Estrogen-related receptor beta-like protein 1; FLJ10147; HIP1 protein interactor; HIP1-interacting protein; huntingtin interacting protein-1 interacting protein; huntingtin-interacting protein-1 protein interactor; intraflagellar transport 57 homolog; Intraflagellar transport protein 57 homolog; MHS4R2; Vestrogen-related receptor beta like 1

          View more View less

          Gene Aliases: 4833420A15Rik; DERP8; ESRRBL1; HIPPI; IFT57; MHS4R2

          View more View less

          UniProt ID: (Human) Q9NWB7, (Mouse) Q8BXG3

          View more View less

          Entrez Gene ID: (Human) 55081, (Dog) 478553, (Mouse) 73916, (Rat) 303968

          View more View less

          Function(s)
          DNA binding protein binding
          Process(es)
          neural tube closure heart looping transcription, DNA-templated regulation of transcription, DNA-templated apoptotic process activation of cysteine-type endopeptidase activity involved in apoptotic process smoothened signaling pathway intraciliary transport cilium assembly regulation of apoptotic process negative regulation of epithelial cell proliferation left/right pattern formation cilium morphogenesis
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-67f4cd694f-jv9gd:80/100.66.135.11:80.
          git-commit: 961b7666e63bcc533d6299fdd217d6a70bd96cf1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0