Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSDQIKFIMD SLNKEPFRKN YNLITFDSLE PMQLLQVLSD VLAEIDPKQL VDIREEMPEQ TAKRMLSLLG ILKYKPSGNA TDMSTFRQGL VIGSKPVIYP VLHWLLQRTN ELKKRAYLAR FLIKLEVPSE FLQDETVADT NKQYEELMEA FKTLHKEYEQ LKISGFSTAE IRKDISAMEE EKDQLIKRVE HLKKRVETAQ NHQWMLKIAR QLRVEKEREE YLAQQKQEQK NQLFHAVQRL QRVQNQLKSM RQAAADAKPE SLMKRLEEEI KFNLYMVTEK FPKELENKKK ELHFLQKVVS EPAMGHSDLL ELESKINEIN TEINQLIEKK MMRNEPIEGK LSLYRQQASI ISRKKEAKAE ELQEAKEKLA SLEREASVKR NQTREFDGTE VLKGDEFKRY VNKLRSKSTV FKKKHQIIAE LKAEFGLLQR TEELLKQRHE NIQQQLQTME EKKGISGYSY TQEELERVSA LKSEVDEMKG RTLDDMSEMV KKLYSLVSEK KSALASVIKE LRQLRQKYQE LTQECDEKKS QYDSCAAGLE SNRSKLEQEV RRLREECLQE ESRYHYTNCM IKNLEVQLRR ATDEMKAYIS SDQQEKRKAI REQYTKNTAE QENLGKKLRE KQKVIRESHG PNMKQAKMWR DLEQLMECKK QCFLKQQSQT SIGQVIQEGG EDRLIL
Isoform CDV-1 may be involved in cardiac hypertrophy caused by carnitine deficiency. Isoform CDV-1R appears to play a role in development of the testis and spermatogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: carnitine deficiency-associated gene expressed in ventricle 1; Carnitine deficiency-associated protein expressed in ventricle 1; CDV-1; intraflagellar transport 81 homolog; Intraflagellar transport protein 81 homolog
Gene Aliases: CDV-1; CDV-1R; CDV1; CDV1R; DV1; IFT81
UniProt ID: (Human) Q8WYA0
Entrez Gene ID: (Human) 28981
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support