Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • IFT88 Antibodies

          Invitrogen

          IFT88 Recombinant Rabbit Monoclonal Antibody (6P4S5)

          View all (10) IFT88 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite IFT88 Recombinant Rabbit Monoclonal Antibody (6P4S5)

          • Antibody Testing Data (1)
          IFT88 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          IFT88 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          IFT88 Antibody (MA5-55471) in WB

          Western Blot analysis of various lysates using IFT88 Recombinant Monoclonal Antibody (Product # MA5-55471) at 1:1,000 dilution. Secondary antibody: HRP-conjugated Goat anti-Rabbit IgG (H+L) at 1:10,000 dilution. Lysates/proteins: 25 μg per lane. Blocking buffer: 3% nonfat dry milk in TBST. Detection: ECL Basic Kit. Exposure time: 180s. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          IFT88 Antibody in Western Blot (WB)

          Product Details

          MA5-55471

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          6P4S5

          Immunogen

          Recombinant fusion protein containing a sequence corresponding to amino acids 534-833 of human IFT88.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.3, with 0.05% BSA, 50% glycerol

          Contains

          0.05% ProClin 300

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          RRID

          AB_3667711

          Product Specific Information

          Immunogen Sequence: GLTYEKLNRL DEALDCFLKL HAILRNSAEV LYQIANIYEL MENPSQAIEW LMQVVSVIPT DPQVLSKLGE LYDREGDKSQ AFQYYYESYR YFPCNIEVIE WLGAYYIDTQ FWEKAIQYFE RASLIQPTQV KWQLMVASCF RRSGNYQKAL DTYKDTHRKF PENVECLRFL VRLCTDLGLK DAQEYARKLK RLEKMKEIRE QRIKSGRDGS GGSRGKREGS ASGDSGQNYS ASSKGERLSA RLRALPGTNE PYESSSNKEI DASYVDPLGP QIERPKTAAK KRIDEDDFAD EELGDDLLPE

          Target Information

          This gene encodes a member of the tetratrico peptide repeat family. Mutations of a similar gene in mouse can cause polycystic kidney disease. Two transcript variants encoding distinct isoforms have been identified for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: HGNC:20606; intraflagellar transport 88 homolog; Intraflagellar transport protein 88 homolog; MGC26259; polaris homolog; probe hTg737 (polycystic kidney disease, autosomal recessive); Recessive polycystic kidney disease protein Tg737; Recessive polycystic kidney disease protein Tg737 homolog; RP11-172H24.2; testicular tissue protein Li 93; tetratricopeptide repeat domain 10; Tetratricopeptide repeat protein 10; Tg737 protein; TgN(Imorpk)737Rpw; TPR repeat protein 10; transgene insert site 737, insertional mutation, polycystic kidney disease

          View more View less

          Gene Aliases: AW552028; D13S1056E; DAF19; flexo; fxo; hTg737; IFT88; orpk; polaris; TG737; Tg737Rpw; TgN737Rpw; TTC10

          View more View less

          UniProt ID: (Human) Q13099, (Mouse) Q61371

          View more View less

          Entrez Gene ID: (Human) 8100, (Mouse) 21821, (Rat) 305918

          View more View less

          Function(s)
          protein binding kinesin binding
          Process(es)
          cilium morphogenesis regulation of cilium assembly regulation of autophagosome assembly eye development kidney development liver development epithelial cell morphogenesis Notch signaling pathway smoothened signaling pathway sperm axoneme assembly spermatid nucleus elongation determination of left/right symmetry nervous system development brain development heart development protein localization epidermis development regulation of smoothened signaling pathway organ morphogenesis anterior/posterior pattern specification dorsal/ventral pattern formation spinal cord dorsal/ventral patterning telencephalon development cell projection organization lung development pancreas development cytoplasmic microtubule organization response to fluid shear stress nonmotile primary cilium assembly epidermal stem cell homeostasis cilium assembly regulation of odontogenesis of dentin-containing tooth embryonic digit morphogenesis positive regulation of insulin-like growth factor receptor signaling pathway regulation of fat cell differentiation forebrain morphogenesis negative regulation of epithelial cell proliferation cardiac muscle cell differentiation palate development inner ear receptor stereocilium organization limb development regulation of feeding behavior cardiac septum morphogenesis lung vasculature development heart formation neural precursor cell proliferation regulation of protein processing cochlea development
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0