Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • IL-29 Antibodies

          Proteintech

          IL-29 Monoclonal Antibody (1H3A3)

          View all (14) IL-29 antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite IL-29 Monoclonal Antibody (1H3A3)

          • Antibody Testing Data (2)
          IL-29 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          IL-29 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          IL-29 Antibody (66163-1-IG) in WB

          Transfected HEK-293 cells were subjected to SDS PAGE followed by western blot with 66163-1-IG ( IL-29 antibody at dilution of 1:5000 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          IL-29 Antibody in Western Blot (WB)
          IL-29 Antibody in Western Blot (WB)

          Product Details

          66163-1-IG

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:5,000-1:50,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          1H3A3

          Immunogen

          IL-29 Fusion Protein Ag12444 (24-200 aa encoded by BC074985)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.21 mg/mL

          Purification

          Protein G

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: TSKPTTTGK GCHIGRFKSL SPQELASFKK ARDALEESLK LKNWSCSSPV FPGNWDLRLL QVRERPVALE AELALTLKVL EAAAGPALED VLDQPLHTLH HILSQLQACI QPQPTAGPRP RGRLHHWLHR LQEAPKKESA GCLEASVTFN LFRLLTRDLK YVADGNLCLR TSTHPEST (24-200 aa encoded by BC074985 )

          Target Information

          IFN-lambdas constitute a novel family of interferons that mediate the induction of anti-viral protection in a wide variety of cells. There are three members in this family, namely lambda1, lambda2, and lambda3, which are also known as IL-29, IL-28A, and IL-28B, respectively. Similar to type I IFNs, IFN-lambdas activate an intracellular signaling to promote gene expression. IFN-lambdas mediate their anti-viral protection through a class II cytokine receptor complex composed of two essential receptor proteins, CRF2-12/IFN-lambdaR1, which is unique to IFN-lambdas, and CFR2-4/IL-10R2, which is shared with IL-10, IL-22, and IL-26 receptors. The IFN-lambdas, IL-28 and IL-29, have recently been reported to prime dendritic cells to induce proliferation of Foxp3-bearing regulatory T cells. DCs that differentiate in response to IFN-lambdas express high levels of class I and II MHC gene products, but low levels of costimulatory molecules. Moreover, these cells can specifically induce IL-2-dependent proliferation of CD4+CD25+FOXP3+ T cells with contact-dependent suppressive activity.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Cytokine Zcyto21; FNL1; IFN lambda1; IFN-lambda-1; IL-29; ILN; Interferon lambda-1; Interferon lambda1; interferon, lambda 1; interferon-lambda-1; Interleukin; interleukin 29 (interferon, lambda 1); Interleukin-29

          View more View less

          Gene Aliases: IFNL1; IL-29; IL29; ZCYTO21

          View more View less

          UniProt ID: (Human) Q8IU54

          View more View less

          Entrez Gene ID: (Human) 282618

          View more View less

          Function(s)
          receptor binding cytokine activity interleukin-28 receptor binding
          Process(es)
          negative regulation of type 2 immune response JAK-STAT cascade negative regulation of cell proliferation negative regulation of interleukin-13 production negative regulation of interleukin-5 production positive regulation of interferon-gamma production positive regulation of tyrosine phosphorylation of STAT protein negative regulation of memory T cell differentiation innate immune response positive regulation of MHC class I biosynthetic process negative regulation of T cell differentiation negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of JAK-STAT cascade positive regulation of immune response defense response to virus
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5d94444789-k2m5x:80/100.66.128.182:80.
          git-commit: ec3573d8beaf0fd412f2d0ea2dcb8dd14c74098c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0