Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MSSSCSGLSR VLVAVATALV SASSPCPQAW GPPGVQYGQP GRSVKLCCPG VTAGDPVSWF RDGEPKLLQG PDSGLGHELV LAQADSTDEG TYICQTLDGA LGGTVTLQLG YPPARPVVSC QAADYENFSC TWSPSQISGL PTRYLTSYRK KTVLGADSQR RSPSTGPWPC PQDPLGAARC VVHGAEFWSQ YRINVTEVNP LGASTRLLDV SLQSILRPDP PQGLRVESVP GYPRRLRASW TYPASWPCQP HFLLKFRLQY RPAQHPAWST VEPAGLEEVI TDAVAGLPHA VRVSARDFLD AGTWSTWSPE AWGTPSTGTI PKEIPAWGQL HTQPEVEPQV DSPAPPRPSL QPHPRLLDHR DSVEQVAVLA SLGTLSFLGL VAGALALGLW LRLRRGGKDG SPKPGFLASV IPVDRRPGAP NL
Interleukin 11 is a stromal cell-derived cytokine that belongs to a family of pleiotropic and redundant cytokines that use the gp130 transducing subunit in their high affinity receptors. This gene encodes the IL-11 receptor, which is a member of the hematopoietic cytokine receptor family. This particular receptor is very similar to ciliary neurotrophic factor, since both contain an extracellular region with a 2-domain structure composed of an immunoglobulin-like domain and a cytokine receptor-like domain. Multiple alternatively spliced transcript variants have been found for this gene. [provided by RefSeq, Jun 2012].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IL-11 receptor subunit alpha; IL-11R subunit alpha; interleukin 11 receptor, alpha; interleukin-11 receptor alpha chain; Interleukin-11 receptor subunit alpha; sIL11RA
Gene Aliases: CRSDA; IL11RA
UniProt ID: (Human) Q14626
Entrez Gene ID: (Human) 3590
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support