Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEPLVTWVVP LLFLFLLPRQ GAACRTSECC FQDPPYPDAD SGSASGPRDL RCYRISSDRY ECSWQYEGPT AGVSHFLRCC LSSGRCCYFA AGSATRLQFS DQAGVSVLYT VTLWVESWAR NQTEKSPEVT LQLYNSVKYE PPLGDIKVSK LAGQLRMEWE TPDNQVGAEV QFRHRTPSSP WKLGDCGPQD DDTESCLCPL EMNVAQEFQL RRRRLGSQGS SWSKWSSPVC VPPENPPQPQ VRFSVEQLGQ DGRRRLTLKE QPTQLELPEG CQGLAPGTEV TYRLQLHMLS CPCKAKATRT LHLGKMPYLS GAAYNVAVIS SNQFGPGLNQ TWHIPADTHT DGMISAHCNL RLPDSRDSPA SASRVAGITG ICHHTRLILY F
The protein encoded by this gene is a type I transmembrane protein that belongs to the hemopoietin receptor superfamily. This protein binds to interleukin 12 with a low affinity, and is thought to be a part of IL12 receptor complex. This protein forms a disulfide-linked oligomer, which is required for its IL12 binding activity. The coexpression of this and IL12RB2 proteins was shown to lead to the formation of high-affinity IL12 binding sites and reconstitution of IL12 dependent signaling. The lack of expression of this gene was found to result in the immunodeficiency of patients with severe mycobacterial and Salmonella infections. Two alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD212; cluster of differentiation 212; IL-12 receptor beta component; IL-12 receptor subunit beta-1; IL-12R subunit beta-1; IL-12RB1; interleukin 12 receptor, beta 1; interleukin-12 receptor beta-1 chain; Interleukin-12 receptor subunit beta-1; MGC34454
Gene Aliases: CD212; IL-12R-BETA1; IL12R; IL12RB; IL12RB1; IMD30
UniProt ID: (Human) P42701
Entrez Gene ID: (Human) 3594
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support