Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The protein encoded by this gene is a receptor for interleukin 20 (IL20), a cytokine that may be involved in epidermal function. The receptor of IL20 is a heterodimeric receptor complex consisting of this protein and interleukin 20 receptor beta (IL20B). This gene and IL20B are highly expressed in skin. The expression of both genes is found to be upregulated in Psoriasis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: class II cytokine receptor ZCYTOR7; CRF2-8; Cytokine receptor class-II member 8; Cytokine receptor family 2 member 8; IL-20 receptor subunit alpha; IL-20R1; interleukin 20 receptor alpha subunit; interleukin 20 receptor, alpha; interleukin-20 receptor I; Interleukin-20 receptor subunit alpha; PRO1315; UNQ681; ZcytoR7
Gene Aliases: CRF2-8; IL-20R-alpha; IL-20R1; IL-20RA; IL20RA; UNQ681/PRO1315
UniProt ID: (Human) B4DLR5
Entrez Gene ID: (Human) 53832
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support