Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: METPAAAAP AGSLFPSFLL LACGTLVAAL LGAAHRLGLF YQLLHKVDKA SVRHGGENVA AVLRAHGVRF IFTLVGGHIS PLLVACEKLG IRVVDTRHEV TAVFAADAMA RLSGTVGVAA VTAGPGLTNT VTAVKNAQMA QSPILLLGGA ASTLLQNRGA LQAVDQLSLF RPLCKFCVSV RRVRDIVPTL RAAMAAAQSG TPGPVFVELP VDVLYPYFMV QKEMVPAKPP KGLVGRVVSW YLENYLANLF AGAWEPQPEG PLPLDIPQAS PQQVQRCVEI LSRAKRPLMV LGSQALLTPT SADKLRAAVE TLGVPCFLGG MARGLLGRNH PLHIRENRSA ALKKADVIVL AGTVCDFRLS YGRVLSHSSK IIIVDRNREE MLLNSDIFWK PQEAVQGDVG SFVLKLVEGL QGQTWAPDWV EELREADRQK EQTFREKAAM PVAQHLNPVQ VLQLVEETLP DNSILVVDGG DFVGTAAHLV QPRGPLRWLD PGAFGTLGVG AGFALGAKLC RPDAEVWCLF GDGAFGYSLI EFDTFVRHKI PVMALVGNDA GWTQISREQV PSLGSNVACG LAYTDYHKAA MGLGARGLLL SRENEDQVVK VLHDAQQQCR DGHPVVVNIL IGRTDFRDGS IAV (1-632 aa encoded by BC011761)
The protein encoded by this gene shares similarity with several thiamine pyrophosphate-binding proteins identified in bacteria, yeast, and plants. The highest degree of similarity is found with bacterial acetolactate synthases (AHAS), which are enzymes that catalyze the first step in branched-chain amino acid biosynthesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-hydroxyacyl-CoA lyase 1 like; 2-hydroxyacyl-CoA lyase 2; acetolactate synthase homolog; Acetolactate synthase-like protein; ilvB (bacterial acetolactate synthase)-like; IlvB-like protein
Gene Aliases: 209L8; AHAS; HACL1L; HACL2; ILV2H; ILVBL
UniProt ID: (Human) A1L0T0
Entrez Gene ID: (Human) 10994
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support