Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
In Drosophila, neuroblasts divide asymmetrically into another neuroblast at the apical side and a smaller ganglion mother cell on the basal side. Cell polarization is precisely regulated by 2 apically localized multiprotein signaling complexes that are tethered by Inscuteable, which regulates their apical localization.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: inscuteable spindle orientation adaptor protein; Minsc; Protein inscuteable homolog
Gene Aliases: 3830422K02Rik; C730020D23; INSC; RGD1311379
UniProt ID: (Human) Q1MX18, (Mouse) Q3HNM7
Entrez Gene ID: (Human) 387755, (Rat) 293166, (Mouse) 233752
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support