Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Osenses
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
The peptide shares 88% identity with rat and mouse sequences.
Specificity of this antibody: INSP3R1.
Intracellular channel that mediates calcium release from the endoplasmic reticulum following stimulation by inositol 1,4,5-trisphosphate. Involved in the regulation of epithelial secretion of electrolytes and fluid through the interaction with AHCYL1. Plays a role in ER stress-induced apoptosis. Cytoplasmic calcium released from the ER triggers apoptosis by the activation of CaM kinase II, eventually leading to the activation of downstream apoptosis pathways.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Inositol 1,4,5 trisphosphate receptor; inositol 1,4,5-triphosphate receptor, type 1; Inositol 1,4,5-trisphosphate receptor type 1; inositol 1,4,5-trisphosphate receptor, type 1; Inositol 1,4,5-trisphosphate-gated calcium channel ITPR1; IP3 receptor; IP3 receptor isoform 1; IP3R 1; protein phosphatase 1, regulatory subunit 94; SI-SIII-SIIA; Type 1 inositol 1,4,5-trisphosphate receptor; Type 1 InsP3 receptor
Gene Aliases: ACV; CLA4; INSP3R1; IP3R; IP3R1; ITPR1; PPP1R94; SCA15; SCA16; SCA29
UniProt ID: (Human) Q14643
Entrez Gene ID: (Human) 3708
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support