Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: AVTLQRAALK FLAKCRKKKK LFAPWRGLQE LTDARRVELK KRVDDYVRRH LGSPMSDVVS RELHAQAQER LQHYFMGRAL EERAQQHREA LIAQISTNVE QLMK
Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 88%.
This gene was first characterized as part of a cluster of genes located within the human major histocompatibility complex class III region. This gene encodes a nuclear protein that is cleaved by caspase 3 and is implicated in the control of apoptosis. In addition, the protein forms a complex with E1A binding protein p300 and is required for the acetylation of p53 in response to DNA damage. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IQ calmodulin-binding motif-containing protein 1; nephrocystin 5; Nephrocystin-5; p53 and DNA damage-regulated IQ motif protein; PIQ
Gene Aliases: IQCB1; KIAA0036; NPHP5; OK/SW-cl.85; PIQ; SLSN5
UniProt ID: (Human) Q15051
Entrez Gene ID: (Human) 9657
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support