Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MALDILAMAP LYQAPAINRI GPKTDPSKRP ADPLKPLVLS RTKLTTIEAK RIMSILDEAI YKVELVTLLS YVASNREDME GMLGEDVMRA VREHEDLCQV LLENVRCLKE KERQLQEQKE AEEEGWLRDR LLSIELQKSS LSPLMQQIKD STKNVLRLLL SNPQAARLLQ MQTQGRSAEA QNFIDSLIEL RGFLFEKLLT SPMEARDKAQ FLQDISRQNS NNQQIIDTLE KELAERMKNR NAEEELEDLD AVHREEKISL EELRRRHKVL VGEFAQIREE REINSKKRME AEQEMVRMVR AATLIQALWK GYLVRSLLRS KKKRGKGKAK DKEKGKQKGK EKGKGKK
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dynein regulatory complex protein 10; dynein regulatory complex subunit 10; IQ domain-containing protein D
Gene Aliases: 4933433C09Rik; CFAP84; DRC10; IQCD
UniProt ID: (Human) Q96DY2
Entrez Gene ID: (Human) 115811
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support