Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MERRAAGPGW AAYERLTAEE MDEQRRQNVA YQYLCRLEEA KRWMEACLKE ELPSPVELEE SLRNGVLLAK LGHCFAPSVV PLKKIYDVEQ LRYQATGLHF RHTDNINFWL SAIAHIGLPS TFFPETTDIY DKKNMPRVVY CIHALSLFLF RLGLAPQIHD LYGKVKFTAE ELSNMASELA KYGLQLPAFS KIGGILANEL SVDEAAVHAA VLAINEAVER GVVEDTLAAL QNPSALLENL REPLAAVYQE MLAQAKMEKA ANARNHDDRE SQDIYDHYLT QAEIQGNINH VNVHGALEVV DDALERQSPE ALLKALQDPA LALRGVRRDF ADWYLEQLNS DREQKAQELG LVELLEKEEV QAGVAAANTK GDQEQAMLHA VQRINKAIRR GVAADTVKEL MCPEAQLPPV YPVASSMYQL ELAVLQQQQG ELGQEELFVA VEMLSAVVLI NRALEARDAS GFWSSLVNPA TGLAEVEGEN AQRYFDALLK LRQERGMGED FLSWNDLQAT VSQVNAQTQE ETDRVLAVSL INEALDKGSP EKTLSALLLP AAGLDDVSLP VAPRYHLLLV AAKRQKAQVT GDPGAVLWLE EIRQGVVRAN QDTNTAQRMA LGVAAINQAI KEGKAAQTER VLRNPAVALR GVVPDCANGY QRALESAMAK KQRPADTAFW VQHDMKDGTA YYFHLQTFQG IWEQPPGCPL NTSHLTREEI QSAVTKVTAA YDRQQLWKAN VGFVIQLQAR LRGFLVRQKF AEHSHFLRTW LPAVIKIQAH WRGYRQRKIY LEWLQYFKAN LDAIIKIQAW ARMWAARRQY LRRLHYFQKN VNSIVKIQAF FRARKAQDDY RILVHAPHPP LSVVRRFAHL LNQSQQDFLA EAELLKLQEE VVRKIRSNQQ LEQDLNIMDI KIGLLVKNRI TLQEVVSHCK KLTKRNKEQL SDMMVLDKQK GLKSLSKEKR QKLEAYQHLF YLLQTQPIYL AKLIFQMPQN KTTKFMEAVI FSLYNYASSR REAYLLLQLF KTALQEEIKS KVEQPQDVVT GNPTVVRLVV RFYRNGRGQS ALQEILGKVI QDVLEDKVLS VHTDPVHLYK NWINQTEAQT GQRSHLPYDV TPEQALSHPE VQRRLDIALR NLLAMTDKFL LAITSSVDQI PYGMRYVAKV LKATLAEKFP DATDSEVYKV VGNLLYYRFL NPAVVAPDAF DIVAMAAGGA LAAPQRHALG AVAQLLQHAA AGKAFSGQSQ HLRVLNDYLE ETHLKFRKFI HRACQVPEPE ERFAVDEYSD MVAVAKPMVY ITVGELVNTH RLLLEHQDCI APDHQDPLHE LLEDLGELPT IPDLIGESIA ADGHTDLSKL EVSLTLTNKF EGLEADADDS NTRSLLLSTK QLLADIIQFH PGDTLKEILS LSASREQEAA HKQLMSRRQA CTAQTPEPLR RHRSLTAHSL LPLAEKQRRV LRNLRRLEAL GLVSARNGYQ GLVDELAKDI RNQHRHRHRR KAELVKLQAT LQGLSTKTTF YEEQGDYYSQ YIRACLDHLA PDSKSSGKGK KQPSLHYTAA QLLEKGVLVE IEDLPASHFR NVIFDITPGD EAGKFEVNAK FLGVDMERFQ LHYQDLLQLQ YEGVAVMKLF NKAKVNVNLL IFLLNKKFLR K
IQGAP3 (IQ motif containing GTPase activating protein 3) is a 1,631 amino acid protein that acts as an effector of Cdc42 and Rac 1, linking their activation to the cytoskeleton during neuronal morphogenesis. A novel member of the IQGAP family, IQGAP3 is highly expressed in brain where it localizes to axons of hippocampal neurons. IQGAP3 contains one Ras-GAP domain, a CH (calponin-homology) domain, four IQ domains and is encoded by a gene located on human chromosome 1, which spans 260 million base pairs, contains over 3,000 genes and comprises nearly 8% of the human genome. Chromosome 1 houses a large number of disease-associated genes, including those that are involved in familial adenomatous polyposis, Stickler syndrome, Parkinson's disease, Gaucher disease, schizophrenia and Usher syndrome. Aberrations in chromosome 1 are found in a variety of cancers, including head and neck cancer, malignant melanoma and multiple myeloma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IQGA3; MGC10170; Ras GTPase-activating-like protein IQGAP3
Gene Aliases: IQGAP3
UniProt ID: (Human) Q86VI3
Entrez Gene ID: (Human) 128239
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support