Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This antibody detects endogenous protein at a molecular weight of 47 kDa.
Purity is >95% by SDS-PAGE.
The IRXA group includes Irx1, Irx2 and Irx4; the IRXB group is comprises Irx3, Irx5 and Irx6. Irx1 and Irx2 are both widely expressed during development in the lung epithelium and also in the ventricular septum. Irx1 and Irx2 also play a role in digit formation (E11.5-E14.5). The Irx gene family members are each expressed in a distinct pattern during mouse heart development. Specifically, Irx1 and Irx2 are expressed in the ventricular septum and Irx3 is expressed in the ventricular trabeculated myocardium. In addition, Irx4 is expressed in the linear heart tube and the AV canal;.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Homeodomain protein IRXA1; Iroquois homeobox protein 1; Iroquois related homeobox 1; Iroquois-class homeodomain protein IRX-1
Gene Aliases: IRX-5; IRX1; IRXA1
UniProt ID: (Human) P78414, (Mouse) P81068
Entrez Gene ID: (Human) 79192, (Rat) 306659, (Mouse) 16371
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support