Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAAAGRLPSS WALFSPLLAG LALLGVGPVP ARALHNVTAE LFGAEAWGTL AAFGDLNSDK QTDLFVLRER NDLIVFLADQ NAPYFKPKVK VSFKNHSALI TSVVPGDYDG DSQMDVLLTY LPKNYAKSEL GAVIFWGQNQ TLDPNNMTIL NRTFQDEPLI MDFNGDLIPD IFGITNESNQ PQILLGGNLS WHPALTTTSK MRIPHSHAFI DLTEDFTADL FLTTLNATTS TFQFEIWENL DGNFSVSTIL EKPQNMMVVG QSAFADFDGD GHMDHLLPGC EDKNCQKSTI YLVRSGMKQW VPVLQDFSNK GTLWGFVPFV DEQQPTEIPI PITLHIGDYN MDGYPDALVI LKNTSGSNQQ AFLLENVPCN NASCEEARRM FKVYWELTDL NQIKDAMVAT FFDIYEDGIL DIVVLSKGYT KNDFAIHTLK NNFEADAYFV KVIVLSGLCS NDCPRKITPF GVNQPGPYIM YTTVDANGYL KNGSAGQLSQ SAHLALQLPY NVLGLGRSAN FLDHLYVGIP RPSGEKSIRK QEWTAIIPNS QLIVIPYPHN VPRSWSAKLY LTPSNIVLLT AIALIGVCVF ILAIIGILHW QEKKADDREK RQEAHRFHFD AM
Modulator of T-cell function. Has a protective effect in graft versus host disease model.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Integrin-alpha FG-GAP repeat-containing protein 1; LINKIN; Protein TIP; T-cell immunomodulatory protein
Gene Aliases: 2310047C21Rik; CDA08; ITFG1; LNKN-1; TIP
UniProt ID: (Human) Q8TB96
Entrez Gene ID: (Human) 81533
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support