Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
This gene encodes an alpha integrin. Integrins are heterodimeric integral membrane glycoproteins composed of an alpha chain and a beta chain that mediate cell-cell and cell-matrix adhesion. The protein encoded by this gene, when bound to the beta 1 chain, forms an integrin that is a receptor for VCAM1, cytotactin and osteopontin. Expression of this gene has been found to be upregulated in small cell lung cancers.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: integrin alpha 9; Integrin alpha-9; Integrin alpha-RLC; integrin, alpha 9
Gene Aliases: (alpha)9; 2610002H11Rik; 6720458D17Rik; AI461869; ALPHA-RLC; D130073C02; D9Ertd428e; ITGA4L; ITGA9; Itga9l; RLC
UniProt ID: (Human) Q13797, (Mouse) B8JK39
Entrez Gene ID: (Human) 3680, (Mouse) 104099, (Rat) 685004
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support