Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
ITGB1BP2 may play a role during maturation and/or organization of muscles cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: integrin beta 1 binding protein (melusin) 2; integrin beta 1 binding protein 2; Integrin beta-1-binding protein 2; Melusin; MGC119214
Gene Aliases: CHORDC3; ITGB1BP; ITGB1BP2; MELUSIN; MSTP015; RGD1565015
UniProt ID: (Human) Q9UKP3, (Mouse) Q9R000
Entrez Gene ID: (Human) 26548, (Rat) 317258, (Mouse) 26549
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support