Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • ITGB6 (Integrin beta 6) Antibodies

          Abnova

          ITGB6 Polyclonal Antibody, MaxPab™

          View all (17) ITGB6 (Integrin beta 6) antibodies
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite ITGB6 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          ITGB6 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ITGB6 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ITGB6 Antibody (H00003694-D01P) in WB

          Western Blot analysis of ITGB6 expression in transfected 293T cell line by ITGB6 MaxPab polyclonal antibody. Lane 1: ITGB6 transfected lysate(85.90 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ITGB6 Antibody in Western Blot (WB)

          Product Details

          H00003694-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          ITGB6 (NP_000879.2, 1 a.a. approximately 788 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MGIELLCLFF LFLGRNDHVQ GGCALGGAET CEDCLLIGPQ CAWCAQENFT HPSGVGERCD TPANLLAKGC QLNFIENPVS QVEILKNKPL SVGRQKNSSD IVQIAPQSLI LKLRPGGAQT LQVHVRQTED YPVDLYYLMD LSASMDDDLN TIKELGSRLS KEMSKLTSNF RLGFGSFVEK PVSPFVKTTP EEIANPCSSI PYFCLPTFGF KHILPLTNDA ERFNEIVKNQ KISANIDTPE GGFDAIMQAA VCKEKIGWRN DSLHLLVFVS DADSHFGMDS KLAGIVIPND GLCHLDSKNE YSMSTVLEYP TIGQLIDKLV QNNVLLIFAV TQEQVHLYEN YAKLIPGATV GLLQKDSGNI LQLIISAYEE LRSEVELEVL GDTEGLNLSF TAICNNGTLF QHQKKCSHMK VGDTASFSVT VNIPHCERRS RHIIIKPVGL GDALELLVSP ECNCDCQKEV EVNSSKCHHG NGSFQCGVCA CHPGHMGPRC ECGEDMLSTD SCKEAPDHPS CSGRGDCYCG QCICHLSPYG NIYGPYCQCD NFSCVRHKGL LCGGNGDCDC GECVCRSGWT GEYCNCTTST DSCVSEDGVL CSGRGDCVCG KCVCTNPGAS GPTCERCPTC GDPCNSKRSC IECHLSAAGQ AREECVDKCK LAGATISEEE DFSKDGSVSC SLQGENECLI TFLITTDNEG KTIIHSINEK DCPKPPNIPM IMLGVSLAIL LIGVVLLCIW KLLVSFHDRK EVAKFEAERS KAKWQTGTNP LYRGSTSTFK NVTYKHREKQ KVDLSTDC

          Target Information

          ITGB6 (Integrin alpha-V/beta-6) is a receptor for fibronectin and cytotactin. It recognizes the sequence R-G-D in its ligands. Internalization of integrin alpha-V/beta-6 via clathrin-mediated endocytosis promotes carcinoma cell invasion.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: integrin beta 6; Integrin beta-6; integrin, beta 6

          View more View less

          Gene Aliases: AI1H; ITGB6

          View more View less

          UniProt ID: (Human) P18564

          View more View less

          Entrez Gene ID: (Human) 3694

          View more View less

          Function(s)
          virus receptor activity integrin binding
          Process(es)
          inflammatory response cell adhesion cell-matrix adhesion integrin-mediated signaling pathway extracellular matrix organization viral entry into host cell
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-9267x:80/100.66.134.224:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0