Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total ITGB6.
ITGB6 (Integrin alpha-V/beta-6) is a receptor for fibronectin and cytotactin. It recognizes the sequence R-G-D in its ligands. Internalization of integrin alpha-V/beta-6 via clathrin-mediated endocytosis promotes carcinoma cell invasion.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: integrin beta 6; Integrin beta-6; integrin, beta 6
Gene Aliases: 2210409C20Rik; 4831415H04Rik; AI1H; ITGB6
UniProt ID: (Human) P18564, (Rat) Q6AYF4, (Mouse) Q9Z0T9
Entrez Gene ID: (Human) 3694, (Rat) 311061, (Mouse) 16420
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support