Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MNNFILLEEQ LIKKSQQKRR TSPSNFKVRF FVLTKASLAY FEDRHGKKRT LKGSIELSRI KCVEIVKSDI SIPCHYKYPF QVVHDNYLLY VFAPDRESRQ RWVLALKEET RNNNSLVPKY HPNFWMDGKW RCCSQLEKLA TGCAQYDPTK NASKKPLPPT PEDNRRPLWE PEETVVIALY DYQTNDPQEL ALRRNEEYCL LDSSEIHWWR VQDRNGHEGY VPSSYLVEKS PNNLETYEWY NKSISRDKAE KLLLDTGKEG AFMVRDSRTA GTYTVSVFTK AVVSENNPCI KHYHIKETND NPKRYYVAEK YVFDSIPLLI NYHQHNGGGL VTRLRYPVCF GRQKAPVTAG LRYGKWVIDP SELTFVQEIG SGQFGLVHLG YWLNKDKVAI KTIREGAMSE EDFIEEAEVM MKLSHPKLVQ LYGVCLEQAP ICLVFEFMEH GCLSDYLRTQ RGLFAAETLL GMCLDVCEGM AYLEEACVIH RDLAARNCLV GENQVIKVSD FGMTRFVLDD QYTSSTGTKF PVKWASPEVF SFSRYSSKSD VWSFGVLMWE VFSEGKIPYE NRSNSEVVED ISTGFRLYKP RLASTHVYQI MNHCWKERPE DRPAFSRLLR QLAEIAESGL
ITK is a tyrosine kinase expressed in T-cells which contains both SH2 and SH3 domains. ITK is thought to play a role in T-cell proliferation and differentiation. The Tec family of non-receptor tyrosine kinases is composed of six proteins designated Tec, Itk (also known as Emt or Tsk), Btk (previously known as Atk, BPK or Emb), Bmx, Txk (also known as Rlk) and Dsrc28C. All members of the family contain SH3 and SH2 domains and, with the exception of Txk and Dsrc28C, also contain pleckstrin homology (PH) and Tec homology (TH) domains in their amino termini. Itk associates with CD28 and becomes activated subsequent to CD28 ligation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: homolog of mouse T-cell itk/tsk; IL-2-inducible T cell kinase; IL-2-inducible T-cell kinase; IL2-inducible T-cell kinase; interleukin-2-inducible T cell kinase; Interleukin-2-inducible T-cell kinase; Kinase EMT; MGC126257; MGC126258; T-cell-specific kinase; Tyrosine-protein kinase ITK/TSK; Tyrosine-protein kinase Lyk
Gene Aliases: EMT; ITK; LPFS1; LYK; PSCTK2
UniProt ID: (Human) Q08881
Entrez Gene ID: (Human) 3702
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support