Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MVKISFQPA VAGIKGDKAD KASASAPAPA SATEILLTPA REEQPPQHRS KRGSSVGGVC YLSMGMVVLL MGLVFASVYI YRYFFLAQLA RDNFFRCGVL YEDSLSSQVR TQMELEEDVK IYLDENYERI NVPVPQFGGG DPADIIHDFQ RRGTYLPQTY IIQEEMVVTE HVSDKEALGS FIYHLCNGKD TYRLRRRATR RRINKRGAKN CNAIRHFENT FVVETLICGV V (1-230 aa encoded by BC025742)
Negative regulator of beta amyloid peptide production. May inhibit the processing of APP by blocking its access to alpha-and beta-secretase. Binding to the beta-secretase-cleaved APP C-terminal fragment is negligible, suggesting that ITM2C is a poor gamma-secretase cleavage inhibitor. May play a role in TNF-induced cell death and neuronal differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BRICHOS domain containing 2C; Cerebral protein 14; hRPC.1050_D_4; Integral membrane protein 2C; integral membrane protein 3; Transmembrane protein BRI3
Gene Aliases: BRI3; BRICD2C; E25; E25C; hucep-14; ITM2C; ITM3; NPD018; PSEC0047
UniProt ID: (Human) Q9NQX7
Entrez Gene ID: (Human) 81618
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support