Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Positive test controls include: Mouse brain, Rat brain.
Immunogen sequence: LLSDSESRSR GNVQLEAGED VGQKNHWQKI RTMVNLPVIS PFKKRYAWVQ LAGHTGSFKA AGTSGLILKR CSEPERYCLA RLMADALRGC VPAFHGVVER DGESYLQLQD LLDGFDGPCV LDCKMGVRTY LEEELTKARE RPKLRKDMYK KMLAVDPEAP TEEEHAQRAV TKPRYMQWRE GISSSTTLGF RIEGIKKADG SCSTDFKTTR SREQVLRVFE EFVQGDEEVL RRYLNRLQQI RDTLEVSEFF RRHEVIGSSL LFVHDHCHRA GVWLIDFGKT TPLPDGQILD HRRPWEEGNR EDGYLLGLDN LIGILASLAE R
Regulates inositol phosphate metabolism by phosphorylation of second messenger inositol 1,4,5-trisphosphate to Ins(1,3,4,5)P4. The activity of the inositol 1,4,5-trisphosphate 3-kinase is responsible for regulating the levels of a large number of inositol polyphosphates that are important in cellular signaling. Both calcium/calmodulin and protein phosphorylation mechanisms control its activity. It is also a substrate for the cyclic AMP-dependent protein kinase, calcium/calmodulin- dependent protein kinase II, and protein kinase C in vitro.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 2.7.1.127; inositol 1,4,5-triphosphate 3-kinase; Inositol 1,4,5-trisphosphate 3-kinase A; inositol 145-triphosphate 3-kinase; Inositol-trisphosphate 3-kinase A; insP 3-kinase A; IP3 3 kinase A; IP3 3-kinase A; IP3-kinase A; IP3K A
Gene Aliases: Itpka
UniProt ID: (Mouse) Q8R071, (Rat) P17105
Entrez Gene ID: (Mouse) 228550, (Rat) 81677
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support