Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: HMDFLSDFE MMLQRKKSMS GKRRRNRDGG TFISDADDVV SAMIVKMNEA AEEDRQLNNQ KKPALKKLTL LPAVVMHLKK QDLKETFIDS GVMSAIKEWL SPLPDRSLPA LKIREELLKI LQELPSVSQE TLKHSGIGRA VMYLYKHPKE SRSNKDMAGK LINEWSRPIF GLTSNYKGMT REEREQRDLE QMPQRRRMNS TGGQTPRRDL EKVLTGEEKA LRPGDPGFCA RARVPMPSNK DYVVRPKWNV EMESSRFQAT SKKGISRLDK QMRKFTDIRK KSRSAHAVKI SIEGNKMPL (523-819 aa encoded by BC017012)
Transcription factor which plays a key role in defining the composition of the RNA polymerase II (RNAPII) elongation complex and in modulating the production of mature mRNA transcripts. Acts as an assembly factor to recruit various factors to the RNAPII elongation complex and is recruited to the complex via binding to the transcription elongation factor SUPT6H bound to the C-terminal domain (CTD) of the RNAPII subunit RPB1 (POLR2A). The SUPT6H:IWS1:CTD complex recruits mRNA export factors (ALYREF/THOC4, EXOSC10) as well as histone modifying enzymes (such as SETD2) to ensure proper mRNA splicing, efficient mRNA export and elongation-coupled H3K36 methylation, a signature chromatin mark of active transcription.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: interacts with Spt6; IWS1 homolog; IWS1-like protein; Protein IWS1 homolog
Gene Aliases: 1700069O15Rik; 3000008H23; AL117927; AW214353; IWS1; IWS1L; RGD1304762
UniProt ID: (Human) Q96ST2, (Mouse) Q8C1D8, (Rat) Q3SWT4
Entrez Gene ID: (Human) 55677, (Mouse) 73473, (Rat) 291705
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support