Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bethyl Laboratories
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Fibronectin type III domain-containing protein 5, the precursor of irisin, is a protein that is encoded by the FNDC5 gene. This gene encodes a secreted protein that is released from muscle cells during exercise. The encoded protein may participate in the development of brown fat. Translation of the precursor protein initiates at a non-AUG start codon at a position that is conserved as an AUG start codon in other organisms. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Fibronectin type III domain-containing protein 5; Fibronectin type III repeat-containing protein 2; PeP; Peroxisomal protein
Gene Aliases: 1500001L03Rik; AI836596; C87088; FNDC5; FRCP2; irisin; PeP; Pxp
UniProt ID: (Human) Q8NAU1, (Mouse) Q8K4Z2
Entrez Gene ID: (Human) 252995, (Mouse) 384061
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support