Antibody tested in indirect ELISA
Isofenphos-methyl is a pesticide that belongs to the group of organophosphates. It is used to control pests in a variety of crops, including fruits, vegetables, and grains. Isofenphos-methyl works by inhibiting the activity of acetylcholinesterase, an enzyme that is essential for proper nerve function. This leads to the accumulation of acetylcholine in the nervous system, which can cause a variety of symptoms in pests, including paralysis and death.