Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Endoplasmic reticulum transmembrane protein involved in vesicle-mediated transport, which is required for neutrophil function. Required for vesicle-mediated transport; it is however unclear whether it is involved in early secretory pathway or intracellular protein transport. Acts as a regulator of neutrophil function, probably via its role in vesicle-mediated transport: required for defense against fungal pathogens and for granulocyte colony-stimulating factor (GM-CSF) signaling pathway; possibly by regulating glycosylation and/or targeting of proteins contributing to the viability and migration of neutrophils.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein jagunal homolog 1
Gene Aliases: GL009; JAGN1; SCN6
UniProt ID: (Human) Q8N5M9
Entrez Gene ID: (Human) 84522
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support