Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • K-Ras (G12A Mutant) Antibodies

          Invitrogen

          K-Ras (G12A Mutant) Recombinant Rabbit Monoclonal Antibody (HL1153)

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite K-Ras (G12A Mutant) Recombinant Rabbit Monoclonal Antibody (HL1153)

          • Antibody Testing Data (2)
          K-Ras (G12A Mutant) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          K-Ras (G12A Mutant) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          K-Ras (G12A Mutant) Antibody (MA5-46940) in WB

          Western blot analysis of K-Ras (G12A Mutant) in Various whole cell extracts (30 µg) separated by 12% SDS-PAGE. Membrane was blotted with K-Ras (G12A Mutant) Monoclonal antibody (Product # MA5-46940) using a dilution of 1:500 followed by HRP-conjugated anti-rabbit IgG secondary antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          K-Ras (G12A Mutant) Antibody in Western Blot (WB)
          K-Ras (G12A Mutant) Antibody in Western Blot (WB)

          Product Details

          MA5-46940

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:10,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          HL1153

          Immunogen

          Carrier-protein conjugated synthetic peptide surrounding mutant G12A of human K-Ras. The exact sequence is proprietary.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          2 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS

          Contains

          no preservative

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2938012

          Product Specific Information

          Store as concentrated solution.

          Centrifuge briefly prior to opening vial.

          Target Information

          This gene, a Kirsten ras oncogene homolog from the mammalian ras gene family, encodes a protein that is a member of the small GTPase superfamily|A single amino acid substitution is responsible for an activating mutation|The transforming protein that results is implicated in various malignancies, including lung adenocarcinoma, mucinous adenoma, ductal carcinoma of the pancreas and colorectal carcinoma|Alternative splicing leads to variants encoding two isoforms that differ in the C-terminal region

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ALPS4; C-BAS/HAS; C-H-RAS; C-HA-RAS1; c-K-ras; c-Ki-ras; c-Ki-ras2; casein kinase 1, alpha 1; casein kinase 1, alpha 1-like; casein kinase I alpha S-like; Casein kinase I isoform alpha; Casein kinase I isoform alpha-like; CASP-1; CASP1 nirs variant 1; caspase 1, apoptosis-related cysteine peptidase (interleukin 1, beta, convertase); Caspase-1; CFC2; CK1; CKI-alpha; CKI-alpha-like; clock regulator kinase; CMNS; CTLO; down-regulated in lung cancer; epididymis secretory sperm binding protein Li 77p; H-RASIDX; HAMSV; HRas proto-oncogene, GTPase; HRAS1; ICE; IL-1 beta-converting enzyme; IL-1BC; IL1B-convertase; interleukin 1, beta, convertase; interleukin 1-B converting enzyme; Interleukin-1 beta convertase; Interleukin-1 beta-converting enzyme; K-Ras; K-RAS2A; K-RAS2B; K-RAS4A; K-RAS4B; Ki-Ras; KRAS; KRAS proto-oncogene, GTPase; KRAS1; KRAS2; N-ras; NCMS; NRAS proto-oncogene, GTPase; NRAS1; NS; NS3; NS6; p21ras; p45; RALD; RASH1; RASK2

          View more View less

          Gene Aliases: CASP1; CK1; CK1a; CKIa; CSNK1A1; CSNK1A1L; HEL-S-77p; HLCDGP1; ICE; IL1BC; IL1BCE; P45; PRO2975

          View more View less

          UniProt ID: (Human) P29466, (Human) P48729, (Human) Q8N752

          View more View less

          Entrez Gene ID: (Human) 834, (Human) 1452, (Human) 122011

          View more View less

          Function(s)
          endopeptidase activity cysteine-type endopeptidase activity protein binding cysteine-type endopeptidase activator activity involved in apoptotic process cysteine-type endopeptidase activity involved in apoptotic process protein kinase activity protein serine/threonine kinase activity ATP binding kinase activity
          Process(es)
          response to hypoxia proteolysis apoptotic process activation of cysteine-type endopeptidase activity involved in apoptotic process signal transduction regulation of autophagy protein processing response to lipopolysaccharide interleukin-1 beta production response to ATP regulation of apoptotic process positive regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of interleukin-1 alpha secretion positive regulation of interleukin-1 beta secretion regulation of inflammatory response mitochondrial depolarization membrane hyperpolarization pyroptosis cellular response to mechanical stimulus cellular response to organic substance programmed necrotic cell death toxin transport cell morphogenesis phagocytosis regulation of cell shape Wnt signaling pathway peptidyl-serine phosphorylation protein phosphorylation Golgi organization mitotic nuclear division cell surface receptor signaling pathway positive regulation of proteasomal ubiquitin-dependent protein catabolic process proteasome-mediated ubiquitin-dependent protein catabolic process intermediate filament cytoskeleton organization cell division negative regulation of canonical Wnt signaling pathway regulation of GTP binding beta-catenin destruction complex assembly beta-catenin destruction complex disassembly
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0