Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MNVRRVESIS AQLEEASSTG GFLYAQNSTK RSIKERLMKL LPCSAAKTSS PAIQNSVEDE LEMATVRHRP EALELLEAQS KFTKKELQIL YRGFKNECPS GVVNEETFKE IYSQFFPQGD STTYAHFLFN AFDTDHNGAV SFEDFIKGLS ILLRGTVQEK LNWAFNLYDI NKDGYITKEE MLDIMKAIYD MMGKCTYPVL KEDAPRQHVE TFFQKMDKNK DGVVTIDEFI ESCQKDENIM RSMQLFENVI
KCNIP4 encodes a member of the family of voltage-gated potassium (Kv) channel-interacting proteins (KCNIPs), which belong to the recoverin branch of the EF-hand superfamily. Members of the KCNIP family are small calcium binding proteins. They all have EF-hand-like domains, and differ from each other in the N-terminus. They are integral subunit components of native Kv4 channel complexes. They may regulate A-type currents, and hence neuronal excitability, in response to changes in intracellular calcium. This protein member also interacts with presenilin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A-type potassium channel modulatory protein 4; Calsenilin-like protein; KChIP4; Kv channel interacting protein 4; Kv channel-interacting protein 4; potassium channel interacting protein 4; Potassium channel-interacting protein 4
Gene Aliases: CALP; KCHIP4; KCNIP4
UniProt ID: (Human) Q6PIL6
Entrez Gene ID: (Human) 80333
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support