Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Osenses
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: KCNJ11.
This receptor is controlled by G proteins. Inward rectifier potassium channels are characterized by a greater tendency to allow potassium to flow into the cell rather than out of it. Their voltage dependence is regulated by the concentration of extracellular potassium; as external potassium is raised, the voltage range of the channel opening shifts to more positive voltages. The inward rectification is mainly due to the blockage of outward current by internal magnesium. Can be blocked by extracellular barium. Subunit of ATP-sensitive potassium channels (KATP). Can form cardiac and smooth muscle-type KATP channels with ABCC9. KCNJ11 forms the channel pore while ABCC9 is required for activation and regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AI842722; ATP-sensitive inward rectifier potassium channel 11; AW491124; beta-cell inward rectifier subunit; BIR; IKATP; Inward rectifier K(+) channel Kir6.2; inwardly rectifying potassium channel KIR6.2; IRK11; KATP; KATP channel; MGC133230; potassium channel inwardly rectifing subfamily J member 11; Potassium channel, inwardly rectifying subfamily J member 11; potassium channel, inwardly rectifying subfamily J, member 11; potassium inwardly rectifying channel, subfamily J, member 11; potassium inwardly-rectifying channel subfamily J member 11; potassium inwardly-rectifying channel, subfamily J, member 11
Gene Aliases: BIR; HHF2; IKATP; KCNJ11; KIR6.2; mBIR; MODY13; PHHI; TNDM3
UniProt ID: (Human) Q14654, (Rat) P70673, (Mouse) Q61743
Entrez Gene ID: (Human) 3767, (Rat) 83535, (Mouse) 16514
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support