Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MKFPIETPRK QVNWDPKVAV PAAAPVCQPK SATNGQPPAP APTPTPRLSI SSRATVVARM EGTSQGGLQT VMKWKTVVAI FVVVVVYLVT GGLVFRALEQ PFESSQKNTI ALEKAEFLRD HVCVSPQELE TLIQHALDAD NAGVSPIGNS SNNSSHWDLG SAFFFAGTVI TTIGYGNIAP STEGGKIFCI LYAIFGIPLF GFLLAGIGDQ LGTIFGKSIA RVEKVFRKKQ VSQTKIRVIS TILFILAGCI VFVTIPAVIF KYIEGWTALE SIYFVVVTLT TVGFGDFVAG GNAGINYREW YKPLVWFWIL VGLAYFAAVL SMIGDWLRVL SKKTKEEVGE IKAHAAEWKA NVTAEFRETR RRLSVEIHDK LQRAATIRSM ERRRLGLDQR AHSLDMLSPE KRSVFAALDT GRFKASSQES INNRPNNLRL KGPEQLNKHG QGASEDNIIN KFGSTSRLTK RKNKDLKKTL PEDVQKIYKT FRNYSLDEEK KEEETEKMCN SDNSSTAMLT DCIQQHAELE NGMIPTDTKD REPENNSLLE DRN
KCNK10 is one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. The message for this gene is highly expressed in the kidney and pancreas. This channel is an open rectifier which primarily passes outward current under physiological K+ concentrations. The protein is stimulated strongly by arachidonic acid and to a lesser degree by membrane stretching, intracellular acidification, and general anaesthetics. Three transcript variants have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2P domain potassium channel TREK2; Outward rectifying potassium channel protein TREK-2; Potassium channel subfamily K member 10; potassium channel TREK-2; potassium channel, subfamily K, member 10; potassium channel, two pore domain subfamily K, member 10; protein phosphatase 1, regulatory subunit 97; TREK-2 K(+) channel subunit; TWIK-related K+ channel 2
Gene Aliases: K2p10.1; KCNK10; PPP1R97; TREK-2; TREK2
UniProt ID: (Human) P57789
Entrez Gene ID: (Human) 54207
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support