Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: MVDRGPLLTS AIIFYLAIGA AIFEVLEEPH WKEAKKNYYT QKLHLLKEFP CLGQEGLDKI LEVVSDAAGQ GVAITGNQTF NNWNWPNAMI FAATVITTIG YGNVAPKTPA GRLFCVFYGL FGVPLCLTWI SALGKFFGGR AKRLGQFLTK RGVSLRKAQI TCTVIFIVWG VLVHLVIPPF VFMVTEGWNY IEGLYYSFIT ISTIGFGDFV AGVNPSANYH ALYRYFVELW IYLGLAWLSL FVNWKVSMFV EVHKAIKKRR RRRKESFESS PHSRKALQVK GSTASKDVNI FSFLSKKEET YNDLIKQIGK KAMKTSGGGE TGPGPGLGPQ GGGLPALPPS LVPLVVYSKN RVPTLEEVSQ TLRSKGHVSR SPDEEAVARA PEDSSPAPEV FMNQLDRISE ECEPWDAQDY HPLIFQDASI TFVNTEAGLS DEETSKSSLE DNLAGEESPQ QGAEAKAPLN MGEFPSSSES TFTSTESELS VPYEQLMNEY NKANSPKGT
KCNK5 is one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. KCNK5 is mainly expressed in the cortical distal tubules and collecting ducts of the kidney. The protein is highly sensitive to external pH and this, in combination with its expression pattern, suggests it may play an important role in renal potassium transport.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acid-sensitive potassium channel protein TASK-2; K2P5.1 potassium channel; Potassium channel subfamily K member 5; potassium channel, subfamily K, member 1 (TASK-2); potassium channel, subfamily, member 5 (KCNK5); potassium channel, two pore domain subfamily K, member 5; TWIK-related acid-sensitive K(+) channel 2; TWIK-related acid-sensitive K+ channel 2
Gene Aliases: K2p5.1; KCNK5; KCNK5b; TASK-2; TASK2
UniProt ID: (Human) O95279
Entrez Gene ID: (Human) 8645
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support