Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 5
Positive Control: Jurkat cell lysate, rat ovary tissue lysate, human kidney tissue, human placenta tissue, mouse small intestine tissue, rat brain tissue.
Subcellular Location: Membrane.
This gene encodes one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. This channel protein, considered an open rectifier, is widely expressed. It is stimulated by arachidonic acid, and inhibited by internal acidification and volatile anaesthetics. [provided by RefSeq, Jul 2008].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ12282; Inward rectifying potassium channel protein TWIK-2; K2P6.1 potassium channel; Potassium channel subfamily K member 6; potassium channel, subfamily K, member 6; potassium channel, two pore domain subfamily K, member 6; potassium inwardly-rectifying channel, subfamily K, member 6; TWIK-2 two-pore-domain K+ channel; TWIK-originated similarity sequence; TWIK-originated sodium similarity sequence
Gene Aliases: D7Ertd764e; K2p6.1; KCNK6; KCNK8; TOSS; TWIK-2; TWIK2
UniProt ID: (Human) Q9Y257, (Rat) G3V8R8, (Mouse) Q3TBV4
Entrez Gene ID: (Human) 9424, (Rat) 116491, (Mouse) 52150
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support