Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium voltage-gated channel subfamily V. This protein is essentially present in the brain, and its role might be to inhibit the function of a particular class of outward rectifier potassium channel types.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: kv2.3r; neuronal potassium channel alpha subunit; Neuronal potassium channel alpha subunit HNKA; potassium channel Kv8.1; potassium channel Kv8.1 homolog; potassium channel, subfamily V, member 1; potassium channel, voltage gated modifier subfamily V, member 1; Potassium voltage-gated channel subfamily V member 1; Voltage-gated potassium channel subunit Kv2.3r; Voltage-gated potassium channel subunit Kv8.1
Gene Aliases: 2700023A03Rik; HNKA; KCNB3; KCNV1; KV2.3; KV8.1; vibe
UniProt ID: (Human) Q6PIU1, (Mouse) Q8BZN2, (Rat) P97557
Entrez Gene ID: (Human) 27012, (Mouse) 67498, (Rat) 60326
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support