Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEGHKAEEEV LDVLRLNVGG CIYTARRESL CRFKDSMLAS MFSGRFPLKT DESGACVIDR DGRLFKYLLD YLHGEVQIPT DEQTRIALQE EADYFGIPYP YSLSDHLANE METYSLRSNI ELKKALTDFC DSYGLVCNKP TVWVLHYLNT SGASCESRII GVYATKTDGT DAIEKQLGGR IHSKGIFKRE AGNNVQYIWS YYSVAELKKM MDAFDAWEGK GVSYWRVPHE LIECWTLEER PLLGSLRHMA PIRKRRLITF NEADESVNYK TGPKPVRFLG PSTSTQIKVK NSASVTVSPA SAIQTSAGAT ANRFQSGSRR KAAQRSAPSR ATALVGTGAP GHPQASPGAA SAENGGTHLP PAKVLLSDKK PTPQRVIKLK RTPLCATAPC LPSPTATRQA NSLKPLPGEA ARALGVRTEN GKNKGN
KCTD18 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include protein homooligomerization.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BTB/POZ domain-containing protein KCTD18; potassium channel tetramerisation domain containing 18
Gene Aliases: 6530404F10Rik; KCTD18
UniProt ID: (Human) Q6PI47
Entrez Gene ID: (Human) 130535
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support