Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • KDELR1 Antibodies

          Invitrogen

          KDELR1 Monoclonal Antibody (KR-10), FITC

          View all (3) KDELR1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite KDELR1 Monoclonal Antibody (KR-10), FITC

          • Antibody Testing Data (1)
          KDELR1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          KDELR1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          KDELR1 Antibody (MA5-45150) in WB

          Western Blot analysis of Rat tissue lysate showing detection of KDEL Receptor protein. Load: 15 µg. Blocking: 1.5% BSA for 30 minutes at RT. Samples were incubated with KDEL Receptor monoclonal antibody (Product # MA5-45150) at 1:1,000 for 2 hours at RT, followed by Sheep Anti-Mouse IgG: HRP for 1 hour at RT. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          KDELR1 Antibody in Western Blot (WB)

          Product Details

          MA5-45150

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunohistochemistry (IHC)

          Assay-dependent
          -

          Immunocytochemistry (ICC/IF)

          1:1,000
          -

          Immunoprecipitation (IP)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Bovine, Dog, Chicken, Fruit fly, Hamster, Human, Mouse, Non-human primate, Sheep, Pig, Rabbit, Rat, Xenopus

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          KR-10

          Immunogen

          A 21 residue synthetic peptide (amino acids 192-212) based on the bovine KDEL receptor and the peptide coupled to KLH
          View immunogen

          Conjugate

          FITC FITC FITC
          • APC 
          • PerCP 
          • Request custom conjugation

          Excitation/Emission Max

          498/517 nm View spectra spectra

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein G

          Storage buffer

          9.1mM sodium bicarbonate/PBS, pH 7.2, with 581mM DMSO, 123mM ethanolamine

          Contains

          no preservative

          Storage conditions

          4°C, store in dark

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2931604

          Product Specific Information

          1 µg/mL was sufficient for detection of KDEL receptor in 20 µg monkey Vero cell lysate by colorimetric immunoblot analysis using Goat Anti-Mouse IgG:AP as the secondary.|Detects approximately 25kDa.

          Target Information

          KDELR1 (KDEL Endoplasmic Reticulum Protein Retention Receptor 1) is a Protein Coding gene. Diseases associated with KDELR1 include Syndromic X-Linked Intellectual Disability 94 and Myopathy, Centronuclear, 1. Among its related pathways are Transport to the Golgi and subsequent modification and Activation of cAMP-Dependent PKA. Gene Ontology (GO) annotations related to this gene include ER retention sequence binding and KDEL sequence binding. An important paralog of this gene is KDELR2.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ER lumen protein retaining receptor 1; ER lumen protein-retaining receptor 1; KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 1; KDEL endoplasmic reticulum protein retention receptor 1; KDEL receptor 1; Putative MAPK-activating protein PM23

          View more View less

          Gene Aliases: 8030486F04Rik; AW215843; BOS_17737; ERD2; ERD2.1; HDEL; I79_020931; KDELR; KDELR1; PM23

          View more View less

          UniProt ID: (Human) P24390, (Bovine) P33946, (Mouse) Q99JH8, (Rat) Q569A6

          View more View less

          Entrez Gene ID: (Human) 10945, (Bovine) 618184, (Sheep) 101122047, (Dog) 484406, (Mouse) 68137, (Rat) 361577, (Chinese hamster) 100750912, (Rabbit) 100349892

          View more View less

          Function(s)
          KDEL sequence binding ER retention sequence binding molecular_function
          Process(es)
          protein retention in ER lumen intracellular protein transport ER to Golgi vesicle-mediated transport retrograde vesicle-mediated transport, Golgi to ER transport biological_process protein transport vesicle-mediated transport
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0