Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • LSD1 Antibodies

          NeoBiotechnologies

          KDM1A (Nuclear Marker and Transcription Factor) Monoclonal Antibody (PCRP-KDM1A-1A10)

          View all (47) LSD1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite KDM1A (Nuclear Marker and Transcription Factor) Monoclonal Antibody (PCRP-KDM1A-1A10)

          • Antibody Testing Data (3)
          KDM1A (Nuclear Marker and Transcription Factor) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          KDM1A (Nuclear Marker and Transcription Factor) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          KDM1A (Nuclear Marker and Transcription Factor) Antibody (23028-MSM1-P1) in IHC (P)

          Formalin-fixed, paraffin-embedded human ovarian carcinoma stained with KDM1A Mouse Monoclonal Antibody (PCRP-KDM1A-1A10). HIER: Tris/EDTA, pH9.0, 45 min. 2°C: HRP-polymer, 30 min. DAB, 5 min. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          KDM1A (Nuclear Marker and Transcription Factor) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          KDM1A (Nuclear Marker and Transcription Factor) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          KDM1A (Nuclear Marker and Transcription Factor) Antibody in Flow Cytometry (Flow)

          Product Details

          23028-MSM1-P1

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          2-4 µg/mL
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1-2 µg/mL
          -

          Flow Cytometry (Flow)

          1-2 µg/mL
          -

          Peptide Array (Array)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          PCRP-KDM1A-1A10

          Immunogen

          Recombinant fragment (around aa152-279) of human KDM1A protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          200 µg/mL

          Purification

          Protein A/G

          Storage buffer

          PBS with 0.05% BSA

          Contains

          0.05% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Positive Control:HeLa cells. Ubiquitous tissue expression.

          Cellular Location: Nucleus.

          Target Information

          LSD1 is a histone demethylase that specifically demethylates 'Lys-4' of histone H3, a specific tag for epigenetic transcriptional activation, thereby acting as a corepressor. LSD1 contains a SWIRM domain, a FAD-binding motif, and an amine oxidase domain. This protein is a component of several histone deacetylase complexes, though it silences genes by functioning as a histone demethylase. It acts by oxidizing the substrate by FAD to generate the corresponding imine that is subsequently hydrolyzed. LSD1 demethylates both mono- and tri-methylted 'Lys-4' of histone H3. This protein may play a role in the repression of neuronal genes. Alone, it is unable to demethylate H3 'Lys-4' on nucleosomes and requires the presence of RCOR1/CoREST to achieve such activity. It may also demethylate 'Lys-9' of histone H3, a specific tag for epigenetic transcriptional repression, thereby leading to derepression of androgen receptor target genes. Mutations affecting this gene can result in cleft palate, psychomotor retardation, and distintive facial features.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: [histone H3]-dimethyl-L-lysine(4) FAD-dependent demethylase 1A; amine oxidase (flavin containing) domain 2; BRAF35-HDAC complex protein BHC110; FAD-binding protein BRAF35-HDAC complex, 110 kDa subunit; Flavin-containing amine oxidase domain-containing protein 2; lysine (K)-specific demethylase 1A; lysine-specific histone demethylase 1; Lysine-specific histone demethylase 1A; RP1-184J9.1

          View more View less

          Gene Aliases: AOF2; BHC110; CPRF; KDM1; KDM1A; KIAA0601; LSD1

          View more View less

          UniProt ID: (Human) O60341

          View more View less

          Entrez Gene ID: (Human) 23028

          View more View less

          Function(s)
          RNA polymerase II transcription factor binding p53 binding chromatin binding transcription factor activity, sequence-specific DNA binding histone deacetylase activity protein binding transcription factor binding oxidoreductase activity enzyme binding ligand-dependent nuclear receptor transcription coactivator activity demethylase activity histone demethylase activity histone demethylase activity (H3-K4 specific) histone demethylase activity (H3-K9 specific) histone demethylase activity (H3-dimethyl-K4 specific) telomeric DNA binding MRF binding transcription regulatory region DNA binding flavin adenine dinucleotide binding androgen receptor binding telomeric repeat-containing RNA binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter in utero embryonic development transcription, DNA-templated regulation of transcription from RNA polymerase II promoter protein demethylation blood coagulation cell proliferation regulation of double-strand break repair via homologous recombination regulation of primitive erythrocyte differentiation histone deacetylation pituitary gland development granulocyte differentiation negative regulation of protein binding histone H3-K9 demethylation positive regulation of histone ubiquitination cellular response to UV histone H3-K4 demethylation negative regulation of DNA binding negative regulation of sequence-specific DNA binding transcription factor activity negative regulation of DNA damage response, signal transduction by p53 class mediator positive regulation of erythrocyte differentiation positive regulation of megakaryocyte differentiation negative regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter positive regulation of hormone biosynthetic process positive regulation of sequence-specific DNA binding transcription factor activity negative regulation of histone H3-K4 methylation negative regulation of histone H3-K9 methylation muscle cell development oxidation-reduction process cellular response to gamma radiation negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator regulation of cellular protein localization positive regulation of neural precursor cell proliferation positive regulation of stem cell proliferation
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0