Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPRGSRARGS KRKRSWNTEC PSFPGERPLQ VRRAGLRTAG AAASLSEAWL RCGEGFQNTS GNPSLTAEEK TITEKHLELC PRPKQETTTS KSTSGLTDIT WSSSGSDLSD EDKTLSQLQR DELQFIDWEI DSDRAEASDC DEFEDDEGAV EISDCASCAS NQSLTSDEKL SELPKPSSIE ILEYSSDSEK EDDLENVLLI DSESPHKYHV QFASDARQIM ERLIDPRTKS TETILHTPQK PTAKFPRTPE NSAKKKLLRG GLAERLNGLQ NRERSAISLW RHQCISYQKT LSGRKSGVLT VKILELHEEC AMQVAMCEQL LGSPATSSSQ SVAPRPGAGL KVLFTKETAG YLRGRPQDTV RIFPPWQKLI IPSGSCPVIL NTYFCEKVVA KEDSEKTCEV YCPDIPLPRR SISLAQMFVI KGLTNNSPEI QVVCSGVATT GTAWTHGHKE AKQRIPTSTP LRDSLLDVVE SQGAASWPGA GVRVVVQRVY SLPSRDSTRG QQGASSGHTD PAGTRACLLV QDACGMFGEV HLEFTMSKAR QLEGKSCSLV GMKVLQKVTR GRTAGIFSLI DTLWPPAIPL KTPGRDQPCE EIKTHLPPPA LCYILTAHPN LGQIDIIDED PIYKLYQPPV TRCLRDILQM NDLGTRCSFY ATVIYQKPQL KSLLLLEQRE IWLLVTDVTL QTKEERDPRL PKTLLVYVAP LCVLGSEVLE ALAGAAPHSL FFKDALRDQG RIVCAERTVL LLQKPLLSVV SGASSCELPG PVMLDSLDSA TPVNSICSVQ GTVVGVDEST AFSWPVCDMC GNGRLEQRPE DRGAFSCGDC SRVVTSPVLK RHLQVFLDCR SRPQCRVKVK LLQRSISSLL RFAAGEDGSY EVKSVLGKEV GLLNCFVQSV TAHPTSCIGL EEIELLSAGG ASAEH
Plays a role in DNA double-strand break (DBS) repair via homologous recombination (HR). Serves as a scaffolding protein that helps to promote the recruitment of DNA-processing enzymes like the helicase BLM and recombinase RAD51 to site of DNA damage, and hence contributes to maintain genomic integrity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA repair-scaffolding protein; k0146; Scaffolding protein involved in DNA repair
Gene Aliases: KIAA0146; SPIDR
UniProt ID: (Human) Q14159
Entrez Gene ID: (Human) 23514
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support