Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDASLEKIAD PTLAEMGKNL KEAVKMLEDS QRRTEEENGK KLISGDIPGP LQGSGQDMVS ILQLVQNLMH GDEDEEPQSP RIQNIGEQGH MALLGHSLGA YISTLDKEKL RKLTTRILSD TTLWLCRIFR YENGCAYFHE EEREGLAKIC RLAIHSRYED FVVDGFNVLY NKKPVIYLSA AARPGLGQYL CNQLGLPFPC LCRVPCNTVF GSQHQMDVAF LEKLIKDDIE RGRLPLLLVA NAGTAAVGHT DKIGRLKELC EQYGIWLHVE GVNLATLALG YVSSSVLAAA KCDSMTMTPG PWLGLPAVPA VTLYKHDDPA LTLVAGLTSN KPTDKLRALP LWLSLQYLGL DGFVERIKHA CQLSQRLQES LKEVNYIKIL VEDELSSPVV VFRFFQELPG SDPVFKAVPV PNMTPSGVGR ERHSCDALNR WLGEQLKQLV PASGLTVMDL EAEGTCLRFS PLMTAAVLGT RGEDVDQLVA CIESKLPVLC CTLQLREEFK QEVEATAGLL YVDDPNWSGI GVVRYEHAND DKSSLKSDPE GENIHAGLLK KLNELESDLT FKIGPEYKSM KSCLYVGMAS DNVDAAELVE TIAATAREIE ENSRLLENMT EVVRKGIQEA QVELQKASEE RLLEEGVLRQ IPVVGSVLNW FSPVQALQKG RTFNLTAGSL ESTEPIYVYK AQGAGVTLPP TPSGSRTKQR LPGQKPFKRS LRGSDALSET SSVSHIEDLE KVERLSSGPE QITLEASSTE GHPGAPSPQH TDQTEAFQKG VPHPEDDHSQ VEGPESLR
PDXDC1 (Pyridoxal Dependent Decarboxylase Domain Containing 1) is a Protein Coding gene. Diseases associated with PDXDC1 include Autosomal Recessive Nonsyndromic Deafness 32 and Fetal Akinesia Deformation Sequence 1. Gene Ontology (GO) annotations related to this gene include pyridoxal phosphate binding and carboxy-lyase activity. An important paralog of this gene is HDC. [GeneCards]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: pyridoxal-dependent decarboxylase domain containing 1; Pyridoxal-dependent decarboxylase domain-containing protein 1
Gene Aliases: KIAA0251; LP8165; PDXDC1
UniProt ID: (Human) Q6P996
Entrez Gene ID: (Human) 23042
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support