Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Recommended positive controls: HCT116, mouse brain.
Predicted reactivity: Mouse (86%), Bovine (90%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The KIAA0323 gene, also known as chromosome 18 open reading frame 8, encodes a protein of currently undefined function. Located on chromosome 18 at the 18q21.1 position, KIAA0323 is part of a family of genes that were initially identified through the sequencing of human brain cDNA libraries. Its precise role in cellular processes remains unclear, but like other KIAA proteins, it may be involved in various cellular functions such as signaling pathways, structural organization, and regulatory mechanisms. Further research is required to elucidate the exact biological pathways in which KIAA0323 participates and its potential implications in human health and disease.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KH and NYN domain-containing protein; Protein KHNYN
Gene Aliases: 6330555F21; 9130227C08Rik; AI605259; KHNYN; KIAA0323; mKIAA0323
UniProt ID: (Human) O15037, (Mouse) Q80U38
Entrez Gene ID: (Human) 23351, (Mouse) 219094
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support