Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDSARLSAAK SSPMMETINM CLQYLDVSVL GELVPRLCEL IRSGVGLGTK GGCASVIVSL TTQCPQDLTP YSGKLMSALL SGLTDRNSVI QKSCAFAMGH LVRTSRDSST EKLLQKLNGW YMEKEEPIYK TSCALTIHAI GRYSPDVLKN HAKEVLPLAF LGMHEIADEE KSEKEECNLW TEVWQENVPG SFGGIRLYLQ ELITITQKAL QSQSWKMKAQ GAIAMASIAK QTSSLVPPYL GMILTALLQG LAGRTWAGKE ELLKAIACVV TACSAELEKS VPNQPSTNEI LQAVLKECSK ENVKYKIVAI SCAADILKAT KEDRFQEFSN IVIPLIKKNS LESSGVRTTK NEEENEKEKE LQLEYLLGAF ESLGKAWPRN AETQRCYRQE LCKLMCERLK LSTWKVQLGV LQSMNAFFQG LMLLEEEHAD PEALAEILLE TCKSITYSLE NKTYSSVRTE ALSVIELLLK KLEESKQWEC LTSECRVLLI ESLATMEPDS RPELQEKAAL LKKTLENLE
Ecm29 appears to be one of three proteins which associate significantly with proteasomes. Ecm29 has been shown to enhance the stability of the proteasome. Ecm29 deletion has been shown to produce 26S proteasomes which dissociated in the absence of ATP. It was thus suggested that Ecm29 tethers the regulatory complex to the proteasome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ECM29 homolog, proteasome accessory protein; Ecm29 proteasome adapter and scaffold; FLJ22036; homolog of yeast Ecm29; KIAA1962; Proteasome adapter and scaffold protein ECM29; Proteasome-associated protein ECM29 homolog; RP11-386D8.2
Gene Aliases: ECM29; ECPAS; KIAA0368
UniProt ID: (Human) Q5VYK3
Entrez Gene ID: (Human) 23392
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support