Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MKNNPLYGDL SLEEAMEERK KNPSWTIEEY DKHSLHTNLS GHLKENPNDL RFWLGDMYTP GFDTLLKKEE KQEKHSKFCR M
Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 86%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KIAA1024-like; Major intrinsically disordered NOTCH2-associated receptor 2; Major intrinsically disordered NOTCH2-binding receptor 1-like; Membrane integral NOTCH2-associated receptor 2
Gene Aliases: KIAA1024L; MINAR2
UniProt ID: (Human) P59773
Entrez Gene ID: (Human) 100127206
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support