Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGAAGRQDFL FKAMLTISWL TLTCFPGATS TVAAGCPDQS PELQPWNPGH DQDHHVHIGQ GKTLLLTSSA TVYSIHISEG GKLVIKDHDE PIVLRTRHIL IDNGGELHAG SALCPFQGNF TIILYGRADE GIQPDPYYGL KYIGVGKGGA LELHGQKKLS WTFLNKTLHP GGMAEGGYFF ERSWGHRGVI VHVIDPKSGT VIHSDRFDTY RSKKESERLV QYLNAVPDGR ILSVAVNDEG SRNLDDMARK AMTKLGSKHF LHLGFRHPWS FLTVKGNPSS SVEDHIEYHG HRGSAAARVF KLFQTEHGEY FNVSLSSEWV QDVEWTEWFD HDKVSQTKGG EKISDLWKAH PGKICNRPID IQATTMDGVN LSTEVVYKKG QDYRFACYDR GRACRSYRVR FLCGKPVRPK LTVTIDTNVN STILNLEDNV QSWKPGDTLV IASTDYSMYQ AEEFQVLPCR SCAPNQVKVA GKPMYLHIGE EIDGVDMRAE VGLLSRNIIV MGEMEDKCYP YRNHICNFFD FDTFGGHIKF ALGFKAAHLE GTELKHMGQQ LVGQYPIHFH LAGDVDERGG YDPPTYIRDL SIHHTFSRCV TVHGSNGLLI KDVVGYNSLG HCFFTEDGPE ERNTFDHCLG LLVKSGTLLP SDRDSKMCKM ITEDSYPGYI PKPRQDCNAV STFWMANPNN NLINCAAAGS EETGFWFIFH HVPTGPSVGM YSPGYSEHIP LGKFYNNRAH SNYRAGMIID NGVKTTEASA KDKRPFLSII SARYSPHQDA DPLKPREPAI IRHFIAYKNQ DHGAWLRGGD VWLDSCRFAD NGIGLTLASG GTFPYDDGSK QEIKNSLFVG ESGNVGTEMM DNRIWGPGGL DHSGRTLPIG QNFPIRGIQL YDGPINIQNC TFRKFVALEG RHTSALAFRL NNAWQSCPHN NVTGIAFEDV PITSRVFFGE PGPWFNQLDM DGDKTSVFHD VDGSVSEYPG SYLTKNDKWH SLASKAASGP SG
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cell migration inducing protein, hyaluronan binding; Cell migration-inducing and hyaluronan-binding protein; colon cancer secreted protein 1; Hyaluronan binding protein involved in hyaluronan depolymerization; hyaluronan-binding protein involved in hyaluronan depolymerization; protein KIAA1199
Gene Aliases: CCSP1; CEMIP; HYBID; KIAA1199; TMEM2L
UniProt ID: (Human) Q8WUJ3
Entrez Gene ID: (Human) 57214
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support